Skip to Content
Merck
All Photos(1)

Documents

50838

Sigma-Aldrich

(R)-Mevalonic acid lithium salt

≥93.0% (qNMR)

Synonym(s):

Lithium (R)-3,5-dihydroxy-3-methylpentanoate, Lithium (R)-3,5-dihydroxy-3-methylvalerate, R-MVA-Li

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11LiO4
CAS Number:
Molecular Weight:
154.09
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.25

Assay

≥93.0% (qNMR)

optical activity

[α]/D -26±3°, c = 0.1 in 0.1 M HCl (16 h)

storage temp.

2-8°C

SMILES string

[Li+].C[C@@](O)(CCO)CC([O-])=O

InChI

1S/C6H12O4.Li/c1-6(10,2-3-7)4-5(8)9;/h7,10H,2-4H2,1H3,(H,8,9);/q;+1/p-1/t6-;/m1./s1

InChI key

PVWNXFFXFNEHDZ-FYZOBXCZSA-M

Application

Mevalonic acid may be used as a reference material in assays for the separation or detection of mevalonic acid. Mevalonic acid is a precursor of isopentenyl pyrophosphate (IPP) and the mevalonate pathway which leads to isoprenoid biosynthesis. Mevalonic acid may be used as a substrate to identify and characterize mevalonate kinase(s).

Biochem/physiol Actions

Mevalonic acid, is an intermediate in the mevalonate pathway, producing terpenes and steroids. This function provides treatment options metabolic disorders , R-mevalonate accumulates in patients with the autosomal recessively inherited mevalonic acidurias, an inborn error of cholesterol and nonsterol isoprene biosynthesis.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Irina Buhaescu et al.
Clinical biochemistry, 40(9-10), 575-584 (2007-05-01)
Mevalonate pathway is an important metabolic pathway which plays a key role in multiple cellular processes by synthesizing sterol isoprenoids, such as cholesterol, and non-sterol isoprenoids, such as dolichol, heme-A, isopentenyl tRNA and ubiquinone. While extensively studied in regard with
S M Houten et al.
Cellular and molecular life sciences : CMLS, 60(6), 1118-1134 (2003-07-16)
Mevalonate kinase (MK) is an essential enzyme in the isoprenoid biosynthesis pathway which produces numerous biomolecules (isoprenoids) involved in a variety of cellular processes. The indispensability of MK and isoprenoid biosynthesis for human health is demonstrated by the identification of
Enantiomerically pure phosphonate analogues of cis- and trans-4-hydroxyprolines.
Cornforth, R. H., et al.
Tetrahedron, 18, 1351-1354 (1962)
G Hoffmann et al.
The New England journal of medicine, 314(25), 1610-1614 (1986-06-19)
A two-year-old boy presented with severe failure to thrive, developmental delay, anemia, hepatosplenomegaly, central cataracts, and dysmorphic features. Quantitative analyses of urinary organic acids revealed massive excretion of mevalonic acid, a metabolic precursor of cholesterol and nonsterol isoprenes: 46,000 to
A Scoppola et al.
Kidney international, 51(3), 908-912 (1997-03-01)
Mevalonic acid (mevalonate or MVA), is an obligate precursor in the biosynthetic pathway of cholesterol. It is partially metabolized by the kidneys and its plasma concentrations are an index of endogenous cholesterol synthesis. The aim of the present study was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service