Skip to Content
Merck
All Photos(1)

Key Documents

46125

Sigma-Aldrich

8-Ethoxy-N,N,N′,N′,N″,N″-hexamethylpyrene-1,3,6-trisulfonamide

suitable for fluorescence, ~85% (TLC)

Synonym(s):

8-Ethoxypyrene-1,3,6-tris(dimethylsulfonamide)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H29N3O7S3
CAS Number:
Molecular Weight:
567.70
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

~85% (TLC)

form

solid

solubility

DMSO: soluble

fluorescence

λex 380 nm; λem 468 nm in DMSO
λex 380 nm; λem 520 nm (Reaction product)

suitability

suitable for fluorescence

SMILES string

CCOc1cc(c2ccc3c(cc(c4ccc1c2c34)S(=O)(=O)N(C)C)S(=O)(=O)N(C)C)S(=O)(=O)N(C)C

InChI

1S/C24H29N3O7S3/c1-8-34-19-13-20(35(28,29)25(2)3)16-11-12-18-22(37(32,33)27(6)7)14-21(36(30,31)26(4)5)17-10-9-15(19)23(16)24(17)18/h9-14H,8H2,1-7H3

InChI key

ZZBMLCMOOSUCTA-UHFFFAOYSA-N

Application

8-Ethoxy-N,N,N′,N′,N″,N″-hexamethylpyrene-1,3,6-trisulfonamide is an analog of 8-hydroxy-N,N,N′,N′,N″,N″-hexamethylpyrene-1,3,6-trisulfonamide (HPTA), a photoacid, used to investigate the steps involved in excited-state deprotonation in polar solvents using pump-probe spectroscopy and time correlated single photon counting fluorescence spectroscopy.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Fluorogenic hydrolase substrate

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Charge redistribution and photoacidity: neutral versus cationic photoacids.
Spry DB, Fayer MD.
J. Chem. Phys. , 128, na-na (2008)
Observation of slow charge redistribution preceding excited-state proton transfer.
Spry DB, Fayer MD.
J. Chem. Phys. , 127, na-na (2007)
D B Spry et al.
The journal of physical chemistry. B, 113(30), 10210-10221 (2009-07-04)
Proton transfer in protonated Nafion fuel cell membranes is studied using several pyrene derivative photoacids. Proton transfer in the center of the Nafion nanoscopic water channels is probed with the highly charged photoacid 8-hydroxypyrene-1,3,6-trisulfonate (HPTS). At high hydration levels, both
E. Koller et al.
Appl. Fluoresc. Technol., 1, 15-15 (1989)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service