Skip to Content
Merck
All Photos(1)

Key Documents

44521

Sigma-Aldrich

MTSEA Biotin

solubility: in DMSO, soluble

Synonym(s):

2-((Biotinoyl)amino)ethyl methanethiosulfonate, N-Biotinylaminoethyl methanethiosulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H23N3O4S3
CAS Number:
Molecular Weight:
381.53
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Pricing and availability is not currently available.

shelf life

limited shelf life, expiry date on the label

solubility

DMSO: soluble

storage temp.

−20°C

SMILES string

CS(=O)(=O)SCCNC(=O)CCCC[C@@H]1SC[C@@H]2NC(=O)N[C@H]12

InChI

1S/C13H23N3O4S3/c1-23(19,20)22-7-6-14-11(17)5-3-2-4-10-12-9(8-21-10)15-13(18)16-12/h9-10,12H,2-8H2,1H3,(H,14,17)(H2,15,16,18)/t9-,10-,12-/m0/s1

InChI key

CQPWMXNDGWWWHG-NHCYSSNCSA-N

Looking for similar products? Visit Product Comparison Guide

Application

MTSEA Biotin, a biotinylated methanethiosulfonate ethylamine reagent, is a thiol-reactive reagent used to label cysteine residues in proteins. MTSEA Biotin labeling is useful to study the structure, function and conformation of biomolecules such as transporters and enzymes.

Biochem/physiol Actions

Thiol-reactive reagent for labeling proteins at cysteine residues to study structure and function.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joachim Ramerstorfer et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 31(3), 870-877 (2011-01-21)
GABA(A) receptors mediate the action of many clinically important drugs interacting with different binding sites. For some potential binding sites, no interacting drugs have yet been identified. Here, we established a steric hindrance procedure for the identification of drugs acting
Ana M Pajor et al.
Biochimica et biophysica acta, 1808(6), 1454-1461 (2010-11-16)
Citric acid cycle intermediates, including succinate and citrate, are absorbed across the apical membrane by the NaDC1 Na+/dicarboxylate cotransporter located in the kidney and small intestine. The secondary structure model of NaDC1 contains 11 transmembrane helices (TM). TM7 was shown
Samuel Freyaldenhoven et al.
The Journal of biological chemistry, 287(21), 17801-17811 (2012-03-28)
In heterologous and endogenous expression systems, we studied the role of ERp44 and its complex partner endoplasmic reticulum (ER) oxidase 1-α (Ero1-Lα) in mechanisms regulating disulfide bond formation for serotonin transporter (SERT), an oligomeric glycoprotein. ERp44 is an ER lumenal
M Traebert et al.
The Journal of membrane biology, 180(1), 83-90 (2001-04-04)
We have combined a functional assay, surface labeling and immunocytochemical methods to compare total and surface-exposed renal type IIa Na+/Pi cotransporter protein. The wild-type type cotransporter (NaPi-IIa) and its functionally comparable cysteine mutant S460C were expressed in Xenopus oocytes. S460C

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service