Skip to Content
Merck
All Photos(1)

Key Documents

43708

Sigma-Aldrich

Xanthan from Xanthomonas campestris

Synonym(s):

Xanthan gum from Xanthomonas campestris, Gum xanthan

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25

biological source

Xanthomonas campestris

Quality Level

form

powder

ign. residue

≤15%

loss

≤15% loss on drying

color

white to beige

viscosity

25-70 mPa.s, 1 % in H2O(25 °C)

suitability

conforms to structure for IR spectroscopy

storage temp.

room temp

InChI

1S/C13H10O/c1-3-7-12-10(5-1)9-11-6-2-4-8-13(11)14-12/h1-8H,9H2

InChI key

GJCOSYZMQJWQCA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Xanthan gum is an anionic polysaccharide composed of a β-(1→4)-D-glucopyranose glucan backbone with side chains of (1→3)-α-D-mannopyranose-(2→1)-β-D-glucuronic acid-(4→1)-β-D-mannopyranose on alternating residues. Approximately half of the terminal mannose residues are 4,6-pyruvated while most of the inner mannose residues are 6-acetylated. Its properties make it a useful matrix component for drug delivery systems. It forms stable drug suspensions in aqueous media and soft gels with locust bean gum or guar gum. Xanthan gum mimics the texture of lipids and is used as a control in experiments where signaling pathways initiated by consumption of lipid-containing reagents are investigated in mice.

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service