Skip to Content
Merck
All Photos(1)

Documents

43324

Sigma-Aldrich

Mesobilirubin

≥97.0% (TLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H40N4O6
CAS Number:
Molecular Weight:
588.69
UNSPSC Code:
12352204

Assay

≥97.0% (TLC)

form

powder or crystals

storage temp.

−20°C

InChI

1S/C33H40N4O6/c1-7-20-19(6)32(42)37-27(20)14-25-18(5)23(10-12-31(40)41)29(35-25)15-28-22(9-11-30(38)39)17(4)24(34-28)13-26-16(3)21(8-2)33(43)36-26/h13-14,34-35H,7-12,15H2,1-6H3,(H,36,43)(H,37,42)(H,38,39)(H,40,41)/b26-13-,27-14-

InChI key

HVHKMUMXERBUAN-IFADSCNNSA-N

Biochem/physiol Actions

The linear tetrapyrrole mesobilirubin, a homologue of bilirubin, is a breakdown product of bile pigments.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mesobilirubin
Fischer, H.
Ber. Deutsch. Chem. Ges., 47, 2330-2333 (1914)
M S Stoll et al.
The Biochemical journal, 163(1), 59-101 (1977-04-01)
Brief reduction of bilirubin with dilute sodium amalgam was shown to give chromogens containing both vinyl and ethylidene beta-substituents. Acid-catalysed rearrangement of these chromogens with bilirubin or mesobilirubin and subsequent dehydrogenation gave a range of new violins containing unconjugated ethylidene
Taihei Ito et al.
Frontiers in pharmacology, 4, 50-50 (2013-05-01)
The aims of this study were to produce mesobiliverdin IXα, an analog of anti-inflammatory biliverdin IXα, and to test its ability to enhance rat pancreatic islet yield for allograft transplantation into diabetic recipients. Mesobiliverdin IXα was synthesized from phycocyanobilin derived
Mesobilirubin as a bile component.
Baumgartel, T.
Medizinische Monatsschrift, 3, 675-677 (1949)
Pasupati Mukerjee et al.
BMC biochemistry, 3, 17-17 (2002-06-25)
Aqueous pKa values of unconjugated bilirubin are important determinants of its solubility and transport. Published pKa data on an analog, mesobilirubin-XIIIalpha, studied by 13C-NMR in buffered solutions containing 27 and 64 vol% (C2H3)2SO because of low aqueous solubility of mesobilirubin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service