Skip to Content
Merck
All Photos(2)

Key Documents

42603

Sigma-Aldrich

Lignin Peroxidase

powder, slightly beige, >0.1 U/mg

Synonym(s):

LiP, Ligninase, Peroxidase, lignin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

form

powder

specific activity

>0.1 U/mg

color

slightly beige

shipped in

wet ice

storage temp.

−20°C

Biochem/physiol Actions

Lignin peroxidase is a fungal enzyme which has a key role in the ligninolytic cycle, the process by which the structural component of plant walls, lignin, is degraded.

Epoxide hydrolase is an enantioselective catalyst for the hydrolytic kinetic resolution of expoxides.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Unit Definition

One unit corresponds to the amount of enzyme, which oxidizes 1 μmole 3.4-dimethoxybenzyl alcohol per minute at pH 3.0 and 30 °C

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Nikita A Khlystov et al.
Communications biology, 4(1), 1027-1027 (2021-09-03)
Lignin has significant potential as an abundant and renewable source for commodity chemicals yet remains vastly underutilized. Efforts towards engineering a biochemical route to the valorization of lignin are currently limited by the lack of a suitable heterologous host for
E N Jacobsen
Accounts of chemical research, 33(6), 421-431 (2000-07-13)
The discovery of the metal salen-catalyzed asymmetric ring-opening (ARO) of epoxides is chronicled. A screening approach was adopted for the identification of catalysts for the addition of TMSN(3) to meso-epoxides, and the chiral (salen)CrN(3) complex was identified as optimal. Kinetic
Andrew T Smith et al.
Proceedings of the National Academy of Sciences of the United States of America, 106(38), 16084-16089 (2009-10-07)
The surface oxidation site (Trp-171) in lignin peroxidase (LiP) required for the reaction with veratryl alcohol a high-redox-potential (1.4 V) substrate, was engineered into Coprinus cinereus peroxidase (CiP) by introducing a Trp residue into a heme peroxidase that has similar

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service