Skip to Content
Merck
All Photos(2)

Key Documents

406-05A

Sigma-Aldrich

Human Osteoblasts: HOb, adult

Synonym(s):

Hob cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
NACRES:
NA.81
Product 406-05A is not currently sold in your country. Contact Technical Service

biological source

human bone (normal)

packaging

pkg of 500,000 cells

manufacturer/tradename

Cell Applications, Inc

growth mode

Adherent

karyotype

2n = 46

morphology

osteoblast

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

arthritis; osteoporosis

shipped in

dry ice

storage temp.

−196°C

General description

Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details.

HOb are isolated from fetal or adult human bones. Additionally, osteoblasts from osteoarthritis or rheumatoid arthritis donors are available. Human osteoblasts provide an excellent model system for studies relevant to the skeletal system.

Characterization: Positive for bone mineralization.

Human osteoblasts have been used to:
  • Characterize signaling pathways mediating bone morphogenesis, such as identify human Crossveinless-2 as an inhibitor of BMP (Binnerts, 2009); demonstrate crosstalk between Runx2, Osterix, and NELL-1 (Chen, 2011); identify TWEAK as a cytokine regulating RANTES production, BMP-2-induced differentiation, and RANKL expression in osteoblasts (Ando, 2006)
  • Demonstrate that Kobophenol A enhances proliferation of osteoblasts via activation of the p38 pathway (Kwak, 2013) and clarify the relationship between hypertension and osteoporosis by demonstrating that Ang II activates osteoclasts via RANKL induction (Shimizu, 2008, 2012)
  • Identify adiponectin as an activator of IL-6, IL-8, VEGF and MMPs in endothelial cells and osteoblasts, implicating it in the development of arthritis (Lee, 2014)
  • Show that inhibition of NF-κB prevented the progression of bone loss in periodontitis and promoted the wound healing in bone defects through the inhibition of osteoclasts (Shimizu, 2009)
  • Study active compounds from Uraria crinita for their potential to stimulate bone formation and regeneration (Mao, 2014)
  • Investigate properties of osteosarcoma, including the role of fluid pressure in angiogenesis (Nathan, 2008) and novel oncogenes (Both, 2012); and develop better treatment strategies (Ma, 2011)
  • Develop optimal materials, coatings and drug delivery techniques for orthopedic implants and tissue engineering (Frandsen, 2014; Ni, 2012; Pilia, 2013a, b, 2014; Shiels, 2012; Valente, 2012; Zhang, 2010, 2011)

Cell Line Origin

Bone

Application

model for skeletal system research, signaling pathways, bone morphogenesis, gene expression, cell activation and proliferation, wound healing, bone formation and regeneration, treatment strategies, tissue engineering, in vitro bone models, mineralization, hormone treatments

Components

Basal Medium containing 10% FBS & 10% DMSO

Preparation Note

  • 2nd passage, >500,000 cells in Basal Medium containing 10% FBS & 10% DMSO
  • Can be cultured at least 10 doublings

Subculture Routine

Please refer to the HOb Culture Protocol.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kiruthika Lawton et al.
International journal of nanomedicine, 14, 7947-7962 (2019-10-22)
Hydroxyapatite (HA) is a biologically active ceramic which promotes bone growth, but it suffers from relatively weak mechanical properties. Multi-walled carbon nanotubes (MWCNTs) have high tensile strength and a degree of stiffness that can be used to strengthen HA; potentially
Taruneet Kaur et al.
Bone, 143, 115603-115603 (2020-08-23)
The study reports a theranostic nature of rno-miR-300 (miR300) in the osteoblast functioning, by influencing the signaling pathway(s), associated with osteoblast differentiation. Excessive expression of miR300 suppresses osteoblast functions. Smad3 served as a validated target for miR300, on homology-based computational

Protocols

Store the cryovials in a liquid nitrogen storage tank immediately upon arrival.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service