Skip to Content
Merck
All Photos(1)

Key Documents

38544

Sigma-Aldrich

O-(Octylphosphoryl)choline solution

1 M in H2O

Synonym(s):

Detergent Screening Solution 33/Kit-No 66317, Phosphocholine octyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7OP(O-)(O)OCH2CH2N+(CH3)3
CAS Number:
Molecular Weight:
295.36
MDL number:
UNSPSC Code:
12161902
PubChem Substance ID:
NACRES:
NA.26

description

zwitterionic

mol wt

295.36 g/mol

concentration

1 M in H2O

technique(s)

protein quantification: suitable

lipid type

phospholipids

storage temp.

2-8°C

SMILES string

CCCCCCCCOP([O-])(=O)OCC[N+](C)(C)C

InChI

1S/C13H30NO4P/c1-5-6-7-8-9-10-12-17-19(15,16)18-13-11-14(2,3)4/h5-13H2,1-4H3

InChI key

MDNHUELOANFCGT-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bindings of Ca2+ ion and monodispersed n-alkylphosphorylcholines to phospholipase A2-II of A. halys blomhoffii.
K Ikeda et al.
Journal of biochemistry, 89(4), 1175-1184 (1981-04-01)
Bindings of monodispersed n-alkylphosphorylcholines to cobra venom phospholipases A2.
K Teshima et al.
Journal of biochemistry, 89(4), 1163-1174 (1981-04-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service