Skip to Content
Merck
All Photos(1)

Key Documents

364M-1

Sigma-Aldrich

Varicella Zoster Virus (SG1-1, SG1-SG4, NCP-1 & IE-62) Mouse Monoclonal Antibody

Sign Into View Organizational & Contract Pricing

Select a Size

364M-17
€155.00
364M-14
€223.00
364M-18
€483.00
364M-15
€502.00
364M-16
€753.00

€155.00


Please contact Customer Service for Availability


Select a Size

Change View
364M-17
€155.00
364M-14
€223.00
364M-18
€483.00
364M-15
€502.00
364M-16
€753.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€155.00


Please contact Customer Service for Availability

Quality Level

100
300
500

biological source

mouse

antibody form

culture supernatant

antibody product type

primary antibodies

clone

SG1-1, SG1-SG4, NCP-1 & IE-62, monoclonal

shipped in

wet ice

storage temp.

2-8°C

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

Product not available in all regions or countries. Analyte Specific Reagent. Analytical and performance characteristics are not established. To request more information on this product, please contact technical services at 800-665-7284 or email: [email protected]

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service