Skip to Content
Merck
All Photos(1)

Key Documents

324M-1

Sigma-Aldrich

PSA (ER-PR8) Mouse Monoclonal Antibody

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€116.00
0.1 ML
€169.00
7 ML
€292.00
0.5 ML
€348.00
1 ML
€495.00

€116.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
€116.00
0.1 ML
€169.00
7 ML
€292.00
0.5 ML
€348.00
1 ML
€495.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€116.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

ER-PR8, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (324M-14)
vial of 0.5 mL concentrate (324M-15)
bottle of 1.0 mL predilute (324M-17)
vial of 1.0 mL concentrate (324M-16)
bottle of 7.0 mL predilute (324M-18)

manufacturer/tradename

Cell Marque®

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50-1:200

isotype

IgG1κ

control

prostate

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic

Gene Information

human ... KLK3(354)

General description

Prostate-Specific Antigen (PSA) is a 33 kDa protein primarily produced by the prostatic epithelium and the epithelial lining of the periurethral glands [1]. PSA is strongly expressed in both normal and neoplastic prostatic tissue. Although PSA can be considered prostate-specific, PSA and/or PSA gene expression has been detected at low levels in some extra-prostatic tissues such as normal breast tissue, breast tumors, endometrium, adrenal neoplasms and renal cell carcinomas.[1][2][3] Anti-PSA is most useful in determining the prostatic origin of carcinomas in non-prostate tissues (metastatic disease) using IHC techniques. This product is best used in conjunction with a panel of antibodies as, up to 27% of prostate carcinoma cases (predominantly poorly differentiated carcinomas) can be negative for this marker.[4]

Quality


IVD

IVD

RUO

RUO

Linkage

PSA Positive Control Slides, Product No. 324S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: [email protected]

Legal Information

Cell Marque is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T J Polascik et al.
The Journal of urology, 162(2), 293-306 (1999-07-20)
Many advances have occurred during the last decade in the clinical use of prostate specific antigen (PSA) for detecting, staging and monitoring prostate cancer. We review the clinical usefulness and limitations of serum PSA as a tumor marker of prostate
K A Alanen et al.
Pathology, research and practice, 192(3), 233-237 (1996-03-01)
Immunohistochemical detection of prostate specific antigen (PSA) in metastases of adenocarcinomas is widely used as an aid to identify the prostatic origin of metastatic cells. However, on the one hand, PSA may not be expressed in some poorly differentiated prostatic
U H Stenman et al.
Seminars in cancer biology, 9(2), 83-93 (1999-04-15)
Prostate specific antigen (PSA) is serine protease produced at high concentrations by normal and malignant prostatic epithelium. It is mainly secreted into seminal fluid, where it digests the gel forming after ejaculation. Only minor amounts of PSA leak out into
M Varma et al.
Histopathology, 47(1), 1-16 (2005-06-29)
Varma M & Jasani B (2005) Histopathology47, 1-16 Diagnostic utility of immunohistochemistry in morphologically difficult prostate cancer: review of current literatureImmunohistochemistry is widely used to distinguish prostate cancer from benign mimics and to establish the prostatic origin of poorly differentiated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service