Skip to Content
Merck
All Photos(1)

Key Documents

30727

Sigma-Aldrich

Decyl-β-D-1-thiomaltopyranoside

≥99.0% (TLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H42O10S
CAS Number:
Molecular Weight:
498.63
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:

description

non-ionic

Quality Level

Assay

≥99.0% (TLC)

optical activity

[α]20/D +53.0±3°, c = 0.5% in methanol

mol wt

498.63 g/mol

CMC

0.9 mM (20-25°C)

storage temp.

−20°C

SMILES string

CCCCCCCCCCS[C@@H]1O[C@H](CO)[C@@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@H]1O

InChI

1S/C22H42O10S/c1-2-3-4-5-6-7-8-9-10-33-22-19(29)17(27)20(14(12-24)31-22)32-21-18(28)16(26)15(25)13(11-23)30-21/h13-29H,2-12H2,1H3/t13-,14-,15-,16+,17-,18-,19-,20-,21-,22+/m1/s1

InChI key

YZNNXXWNKQOETJ-HYLFJBCQSA-N

Application

Detergent for the purification, extraction and solubilization of membrane-bound proteins in the native structure. It increases solubilization by dissociating aggregates and is easily removed by dialysis.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mechref, Y. and Rassi, Z.E.
Journal of Chromatography A, 757, 263-263 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service