Skip to Content
Merck

Skip To

28811

Cadmium chloride 0.1 M solution

Synonym(s):

Cadmium chloride solution, Additive Screening Solution 23/Kit-No 78374

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
12352202
PubChem Substance ID:
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


density

1.01 g/mL at 20 °C

storage temp.

2-8°C

SMILES string

Cl[Cd]Cl

InChI

1S/Cd.2ClH/h;2*1H/q+2;;/p-2

InChI key

YKYOUMDCQGMQQO-UHFFFAOYSA-L



signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Carc. 1B - Muta. 1B - Repr. 1B - STOT RE 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

10108-64-2

CAS No.

10108-64-2

CAS No.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



M Yde et al.
Euro surveillance : bulletin Europeen sur les maladies transmissibles = European communicable disease bulletin, 17(38) (2012-10-09)
A cluster of time-linked cases and the identification of a clonal strain suggest the occurrence of an outbreak of listeriosis in Belgium in 2011, presumably due to the consumption of hard cheese made with pasteurised milk and produced by a
Michael D Menz et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 33(10), 4550-4560 (2013-03-08)
Focused ultrasound is a promising noninvasive technology for neural stimulation. Here we use the isolated salamander retina to characterize the effect of ultrasound on an intact neural circuit and compared these effects with those of visual stimulation of the same
Vanessa Cardoso Pires et al.
The British journal of nutrition, 110(11), 2020-2029 (2013-05-10)
The aim of the present study was to investigate the effects of long-term grape juice concentrate (GJC) consumption, in two dosages, on the reproductive parameters of cadmium-exposed male rats. The effects of the concentrate on body mass gain, plasma testosterone



Questions

Reviews

No rating value

Active Filters