Skip to Content
Merck
All Photos(1)

Key Documents

27689

Sigma-Aldrich

Concanamycin A

from Streptomyces sp., ≥80% (HPLC)

Synonym(s):

Folimycin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H75NO14
CAS Number:
Molecular Weight:
866.09
Beilstein:
3560277
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25
To inquire about this product 27689, please contact your local Merck office or dealer. Contact Technical Service

biological source

Streptomyces sp.

Quality Level

Assay

≥80% (HPLC)

form

solid

antibiotic activity spectrum

viruses

Mode of action

enzyme | inhibits

storage temp.

−20°C

SMILES string

CCC1C(O)C(C)C\C(C)=C\C=C\C(OC)C(OC(=O)\C(OC)=C\C(C)=C\C(C)C1O)C(C)C(O)C(C)[C@]2(O)C[C@@H](O[C@H]3C[C@@H](O)[C@H](OC(N)=O)[C@@H](C)O3)[C@H](C)C(O2)\C=C\C

InChI

1S/C46H75NO14/c1-13-16-34-28(7)37(58-38-22-33(48)43(31(10)57-38)60-45(47)53)23-46(54,61-34)30(9)41(51)29(8)42-35(55-11)18-15-17-24(3)19-26(5)39(49)32(14-2)40(50)27(6)20-25(4)21-36(56-12)44(52)59-42/h13,15-18,20-21,26-35,37-43,48-51,54H,14,19,22-23H2,1-12H3,(H2,47,53)/b16-13+,18-15+,24-17+,25-20+,36-21-/t26?,27?,28-,29?,30?,31-,32?,33-,34?,35?,37-,38+,39?,40?,41?,42?,43-,46+/m1/s1

InChI key

DJZCTUVALDDONK-LWLXYWDNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: macrolide

Application

Concanamycin A has been used as an inhibitor of the perforin pathway.[1]

Other Notes

Characterization of concanamycins A, B and C;[2] Selective inhibitor of the vacuolar proton-ATPase.[3][4][5]

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 2 Oral - Eye Irrit. 2

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Concanamycin A, the specific inhibitor of V-ATPases, binds to the V(o) subunit c.
Huss M
The Journal of Biological Chemistry, 277(43), 40544-40548 (2002)
Cytotoxic mechanisms in different forms of T-cell-mediated drug allergies
P. C. Kuechler
Allergy (2004)
R Guinea et al.
FEBS letters, 349(3), 327-330 (1994-08-08)
The selective inhibitor of the vacuolar proton-ATPase, concanamycin A, powerfully blocks influenza virus entry into cells, if present during the initial times of virus infection. Attachment of virus particles to cells is not prevented by concanamycin A, rather the exit
H. Kinashi et al.
The Journal of Antibiotics, 38, 1333-1333 (1984)
Wucheng Tao et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 33(9), 3738-3743 (2013-03-01)
Ambient GABA in the brain tonically activates extrasynaptic GABA(A) receptors, and activity-dependent changes in ambient GABA concentration can also activate GABA(B) receptors. To investigate an interaction between postsynaptic GABA(B) and GABA(A) receptors, we recorded GABA(A) currents elicited by exogenous GABA

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service