Skip to Content
Merck
All Photos(1)

Documents

257R-1

Sigma-Aldrich

GCDFP-15 (EP1582Y) Rabbit Monoclonal Primary Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

EP1582Y, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (257R-14)
vial of 0.5 mL concentrate (257R-15)
bottle of 1.0 mL predilute (257R-17)
vial of 1.0 mL concentrate (257R-16)
bottle of 7.0 mL predilute (257R-18)

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500

isotype

IgG

control

breast, breast carcinoma

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic

Gene Information

human ... PIP(5304)

General description

Gross cystic disease fluid protein-15 (GCDFP-15) is one of four major component proteins found in the cystic fluid obtained from patients with fibrocystic changes of the breast. GCDFP-15 is a marker of apocrine glandular differentiation in both benign and malignant mammary epithelium. This protein has a widespread distribution in apocrine glands elsewhere in the axillary and perianal tissues, as well as in the sublingual and submaxillary salivary glands. The GCDFP-15 protein is a 15 kDa glycoprotein shown to be prolactin-inducible, and the GCDFP-15 gene has been cloned. Ultrastructurally, the GCDFP-15 protein has been localized in Golgi vesicle and cytoplasmic granules. The protein is released by exocytosis at the apices of the mammary epithelial cells. The primary application for this antibody product is the identification of breast carcinoma in the metastatic setting, or fluid analysis. In a study by Wick et al. on paraffin sections of 105 breast cancers and 585 non-mammary malignancies in order to assess its value in this context, anti-GCDFP-15 was compared with another putative mammary epithelial marker, alpha-lactalbumin (ALA), with respect to sensitivity and specificity for a diagnosis of breast carcinoma. Overall, the rates of specificity and sensitivity and positive predictive value for GCDFP-15 were 95%, 74%, and 74%, respectively. A consistent congruency between the reactivity patterns of primary and metastatic breast cancers was noted for GCDFP-15. Besides mammary carcinomas, the major tumor types that express GCDFP-15 were carcinomas of the salivary glands and sweat glands. Since the latter two types of lesions are unlikely to be diagnosed as metastatic breast cancer, statistical indices were recalculated after exclusion of these three tumor types. Following this exclusion, the adjusted rate of specificity of GCDFP-15 and positive predictive value for a diagnosis of metastatic carcinoma of the breast were each 99%. Another study by Bhargava et al. has shown GCDFP-15 staining of a higher percentage of cancer cells in lobular carcinoma as compared to ductal carcinoma of the breast.

Linkage

GCDFP-15 Positive Control Slides, Product No. 257S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Mazoujian et al.
The American journal of pathology, 110(2), 105-112 (1983-02-01)
Gross cystic disease fluid is a pathologic secretion from breast composed of several glycoproteins, including a unique 15,000-dalton monomer protein, GCDFP-15. By the immunoperoxidase technique, GCDFP-15 was localized in the apocrine metaplastic epithelium lining breast cysts and in apocrine glands
S Ansai et al.
The American Journal of dermatopathology, 17(3), 249-255 (1995-06-01)
We investigated immunohistochemically the localization of lysozyme and Leu M1 in normal skin, 76 cases of benign sweat gland tumors, 28 cases of malignant sweat gland tumors, 23 cases of extramammary Paget's disease, 7 cases of sebaceous carcinoma, 6 cases
U Raju et al.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 6(5), 516-520 (1993-09-01)
Signet ring carcinoma of the breast often metastasizes to gastrointestinal tract and female genital tract. We report clinicopathologic features of 10 breast carcinomas with signet ring features, five of which had unusual metastatic patterns. The primary breast tumor in all
G Mazoujian et al.
The American Journal of dermatopathology, 10(1), 28-35 (1988-02-01)
Sixty-five cases of benign sweat gland tumors of the skin were studied for the expression and localization of gross cystic disease fluid protein-15 (GCDFP-15) by immunoperoxidase methods. There was positive staining of tumors of probable apocrine differentiation in 10 of
C Cohen et al.
Archives of pathology & laboratory medicine, 117(3), 291-294 (1993-03-01)
Mammary Paget's disease has been said to result from epidermal spread by contiguity of primary intraductal carcinoma. To assess similar identity, we immunostained Paget's cells and underlying intraductal and/or invasive mammary carcinoma in 20 cases for cytokeratins, epithelial membrane antigen

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service