Skip to Content
Merck
All Photos(2)

Documents

252-05N

Sigma-Aldrich

Human Umbilical Artery Smooth Muscle Cells: HUASMC, neonatal

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
NACRES:
NA.81

biological source

human umbilicus (normal arteries)

Quality Level

packaging

pkg of 500,000 cells

manufacturer/tradename

Cell Applications, Inc

growth mode

Adherent

karyotype

2n = 46

morphology

smooth muscle

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

diabetes; stroke; cardiovascular diseases

shipped in

dry ice

storage temp.

−196°C

Related Categories

General description

Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details.

HUASMC has been used to study effects of snake venom on smooth muscle cells (Mora, 2008); secretion mechanism of the (pro)renin receptor (Yoshikawa, 2011) and the role of proteasomes (Martin, 2014) in the development of cardiovascular diseases; and to develop novel material surfaces to inhibit smooth muscle cell proliferation at the same time allowing endothelial cells adhesion and proliferation in order to reduce risk associated with vascular implants (Kanie, 2012). Characterization: positive for smooth muscle cell specific alpha-actin expression.

Cell Line Origin

Artery

Application

vascular function, tissue engineering, pharmacology of vascular tissue

Components

Smooth Muscle Cell Basal Medium containing 10% FBS & 10% DMSO

Preparation Note

  • 2nd passage, >500,000 cells in Smooth Muscle Cell Basal Medium containing 10% FBS & 10% DMSO
  • Can be cultured at least 16 doublings

Subculture Routine

Please refer to the HUASMC Culture Protocol.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service