Skip to Content
Merck
All Photos(1)

Key Documents

25102

Sigma-Aldrich

9-(Chloromethyl)anthracene

BioReagent, suitable for fluorescence, ≥97.0% (AT)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H11Cl
CAS Number:
Molecular Weight:
226.70
Beilstein:
1873394
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Pricing and availability is not currently available.

product line

BioReagent

Assay

≥97.0% (AT)

mp

138-140 °C (lit.)

fluorescence

λex 356 nm; λem 412 nm (after derivatisation with sodium acetate)

suitability

suitable for fluorescence

SMILES string

ClCc1c2ccccc2cc3ccccc13

InChI

1S/C15H11Cl/c16-10-15-13-7-3-1-5-11(13)9-12-6-2-4-8-14(12)15/h1-9H,10H2

InChI key

PCVRSXXPGXRVEZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

In addition to the emission maximum at 412 nm, there are lower maxima at 434, 395 and 459 nm.

Other Notes

Blocking group reagent for carboxylic acids, phenols, thiophenols and mercaptans[1]; Derivatizing agent for carboxylic acids giving esters with enhanced UV and fluorescence detection in HPLC[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W.D. Korte
Journal of Chromatography A, 243, 153-153 (1982)
Dorothea F K Rawn et al.
Journal of chromatography. A, 1080(2), 148-156 (2005-07-13)
A rapid and simple method for confirmation of the diarrhetic shellfish poisons (DSP): okadaic acid (OA), dinophysistoxin-1 (DTX-1) and dinophysistoxin-2 (DTX-2) using fluorescence detection following derivatization with 9-chloromethylanthracene, has been established as an alternate to LC/MS. Exposure of the anthrylmethyl
Jason P Schrum et al.
Journal of the American Chemical Society, 131(40), 14560-14570 (2009-09-18)
The development of a sequence-general nucleic acid copying system is an essential step in the assembly of a synthetic protocell, an autonomously replicating spatially localized chemical system capable of spontaneous Darwinian evolution. Previously described nonenzymatic template-copying experiments have validated the
Ying Zhang et al.
Colloids and surfaces. B, Biointerfaces, 44(2-3), 104-109 (2005-07-26)
Polymeric micelles based on amphiphilic diblock copolymers methoxy poly(ethylene glycol)-polylactide with various hydrophobic lengths were designed as carriers of poorly water-soluble anticancer drug methotrexate (MTX). Relationship between physicochemical characteristics of micelles and release behavior was explored. The critical micelle concentration
J F Lawrence et al.
Journal of chromatography. A, 721(2), 359-364 (1996-01-19)
The reagent 9-chloromethylanthracene was evaluated for derivatization of the diarrhetic shellfish poisons, okadaic acid and dinophysistoxin-1 (DTX-1), to form fluorescent products separable by liquid chromatography. The toxins were reacted with the reagent in acetonitrile in the presence of tetramethylammonium hydroxide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service