Skip to Content
Merck
All Photos(1)

Key Documents

230R-1

Sigma-Aldrich

Caldesmon (E89) Rabbit Monoclonal Primary Antibody

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€168.00
0.1 ML
€217.00
0.5 ML
€579.00
7 ML
€643.00
1 ML
€889.00

€168.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
€168.00
0.1 ML
€217.00
0.5 ML
€579.00
7 ML
€643.00
1 ML
€889.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€168.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

E89, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (230R-14)
vial of 0.5 mL concentrate (230R-15)
bottle of 1.0 mL predilute (230R-17)
vial of 1.0 mL concentrate (230R-16)
bottle of 7.0 mL predilute (230R-18)

manufacturer/tradename

Cell Marque®

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:25-1:100

isotype

IgG

control

appendix, breast

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic

General description

Caldesmon is a regulatory protein that plays a role in smooth muscle contraction with actin, myosin, tropomyosin, and calmodulin. Anti-caldesmon antibody labels smooth muscle and tumors of smooth muscle.[1][2] It also typically shows expression in epithelioid mesothelioma but not in ovarian serous carcinoma.[3][4]

Quality


IVD

IVD

IVD

RUO

Linkage

Caldesmon Positive Control Slides, Product No. 230S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: [email protected]

Legal Information

Cell Marque is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W Glenn McCluggage
Advances in anatomic pathology, 11(3), 162-171 (2004-04-21)
In recent years there have been a plethora of publications regarding the value of immunohistochemical studies in diagnosis in gynecological pathology. In many instances, papers are published initially that suggest that a certain antibody or panel of antibodies is of
Camilla E Comin et al.
The American journal of surgical pathology, 30(4), 463-469 (2006-04-21)
Although a large number of immunohistochemical markers that can facilitate the differential diagnosis between epithelioid pleural mesothelioma and lung adenocarcinoma involving the pleura have proven to be valuable, no single antibody has demonstrated absolute sensitivity and/or specificity in making this
Markku Miettinen et al.
Archives of pathology & laboratory medicine, 130(10), 1466-1478 (2006-11-09)
Gastrointestinal stromal tumors (GISTs) are specific, generally Kit (CD117)-positive, mesenchymal tumors of the gastrointestinal tract encompassing a majority of tumors previously considered gastrointestinal smooth muscle tumors. They are believed to originate from interstitial cells of Cajal or related stem cells.
K Watanabe et al.
Human pathology, 30(4), 392-396 (1999-04-20)
h-Caldesmon (h-CD) is a protein combined with actin and tropomyosin that regulates cellular contraction. h-CD has been thought to be expressed exclusively in vascular and visceral smooth muscle cells (SMC). We examined h-CD expression immunohistochemically in tumors with SMC and
Camilla E Comin et al.
The American journal of surgical pathology, 31(8), 1139-1148 (2007-08-02)
Distinguishing between epithelioid peritoneal mesothelioma and papillary serous carcinomas involving the peritoneum may be very difficult, owing to overlapping morphologic features. Immunohistochemistry may facilitate establishing a correct diagnosis, but, as no single antibody has demonstrated absolute sensitivity and specificity for

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service