Skip to Content
Merck
All Photos(1)

Key Documents

21872

Sigma-Aldrich

(2-Carboxyethyl)-β-cyclodextrin sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C51H76Na3O41
Molecular Weight:
1414.09
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25

form

solid

optical activity

[α]20/D +129±6°, c = 1% in H2O

impurities

≤10% acidic form
~5% water

solubility

H2O: 50 mg/mL, clear, colorless

storage temp.

room temp

General description

(2-Carboxyethyl)-β-cyclodextrin sodium is a derivative of β-cyclodextrin, typically used to enhance the properties of the latter such as stability and water solubility. β-cyclodextrin has garnered increased research attention due to its potential application as host-guest inclusion complex used in the pharmaceutical industry and as antibacterial material utilized in food and pharmacy.[1]

Application

(2-Carboxyethyl)-β-cyclodextrin sodium salt may be used:
  • As a chiral selector for the enantioseparation of cinacalcet impurities[2] and fenamiphos pesticide[3] by capillary zone electrophoresis and 31P nuclear magnetic resonance spectroscopy (31P NMR), respectively.
  • To form an inclusion complex with carvacrol for subsequent use in the antibacterial multilayer construction with chitosan, loaded onto poly(L-lactic acid) (PLLA).[1]

Cyclodextrins (CD) are cyclic oligosaccharides composed of D-glucose units connected via α(1→4) glycosidic bonds. Common cyclodextrins contain six (α-CD); seven (β-CD) or eight (gamma-CD) D-glucose units. Cyclodextrins are sometimes derivitized through esterification at positions two, three and/or six.(2-Carboxyethyl)-β-cyclodextrin and other negative charge derivitized cyclodextrins are used in chiral recognition and resolution of cationic racemic mixtures of enantiomers using capillary electrophoresis.

Analysis Note

average degree of substitution, DS ~3

Other Notes

To gain a comprehensive understanding of our extensive range of Oligosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Koppenhoefer et al.
Journal of chromatography. A, 875(1-2), 135-161 (2000-06-06)
This is a selected review, highlighting our results obtained in an extended screening program ("The German-Chinese Drug Screening Program"), with a focus on a set of original data obtained with heptakis(2,3,6-tri-O-methyl)-beta-cyclodextrin (TM-beta-CD) as the chiral solvating agent (CSA). The enantioseparation
Benedetta Pasquini et al.
Journal of chromatography. A, 1568, 205-213 (2018-07-15)
A capillary electrophoresis method for the simultaneous determination of the enantiomeric purity and of impurities of the chiral calcimimetic drug cinacalcet hydrochloride has been developed following Quality by Design principles. The scouting phase was aimed to select the separation operative
L J Jin et al.
Journal of chromatography. B, Biomedical sciences and applications, 708(1-2), 257-266 (1998-07-08)
The enantiomeric separation of some racemic anti-histamines and anti-malarials, namely (+/-)-pheniramine, (+/-)-brompheniramine, (+/-)-chlorpheniramine, (+/-)-doxylamine, and (+/-)-chloroquine, was investigated by capillary zone electrophoresis. The enantiomeric separation of five compounds was obtained by addition of approximately 7 mM (1%, w/v) sulfated-beta-cyclodextrin into
Antibacterial multilayer of chitosan and (2-carboxyethyl)-beta-cyclodextrin onto polylactic acid (PLLA)
Andrade-Del O, et al.
Food Hydrocolloids, 88, 228-236 (2019)
E Albers et al.
Critical reviews in therapeutic drug carrier systems, 12(4), 311-337 (1995-01-01)
The current cyclodextrin (CD) literature is reviewed concerning synthesis, characterization, and pharmaceutical relevant applications of CD derivatives. Although natural CDs have been used extensively to improve pharmaceutical properties, the effects of chemically modified CDs on the solubility, dissolution rate, and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service