Skip to Content
Merck
All Photos(1)

Key Documents

179R-1

Sigma-Aldrich

CD79a (SP18) Rabbit Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

SP18, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (179R-14)
vial of 0.5 mL concentrate (179R-15)
bottle of 1.0 mL predilute (179R-17)
vial of 1.0 mL concentrate (179R-16)
bottle of 7.0 mL predilute (179R-18)

manufacturer/tradename

Cell Marque®

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:200-1:1000

isotype

IgG1

control

tonsil

shipped in

wet ice

storage temp.

2-8°C

visualization

membranous

Gene Information

human ... CD79A(973)

General description

Anti-CD79a recognizes the Ig-alpha protein of the B-cell antigen component of the B-lymphocyte antigen receptor.[1][2] Anti-CD79a is generally used to complement CD20. This antibody will stain many of the same lymphomas as CD20, but also is more likely to stain precursor B-cell lymphoblastic lymphoma/leukemia. Anti-CD79a also stains more cases of plasma cell myeloma and occasionally some types of endothelial cells as well.

Linkage

CD79a Positive Control Slides, Product No. 179S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: [email protected]

Legal Information

Cell Marque is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diagnostic Immunohistochemistry: Theranostic and Genomic Applications (2006)
D Y Mason et al.
Blood, 86(4), 1453-1459 (1995-08-15)
The CD79 molecule, comprising two polypeptide chains, mb-1 (CD79a) and B29 (CD79b), is physically associated in the B-cell membrane with immunoglobulin. It transmits a signal after antigen binding and may, therefore, be considered the B cell equivalent of CD3. It
P J Kurtin et al.
American journal of clinical pathology, 112(3), 319-329 (1999-09-09)
The immunoperoxidase technique was used with antibodies against B-cell-associated antigens, including CD20, CD79a, CD10, CD23, CD43, cyclin D1, bcl-2, and kappa and lambda immunoglobulin light chains on formalin-fixed and B5-fixed tissue sections of follicular, small lymphocytic, mantle cell, and marginal

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service