Skip to Content
Merck
All Photos(1)

Documents

14764

Sigma-Aldrich

1-Deoxy-D-xylulose

≥80% (TLC)

Synonym(s):

(3S,4R)-3,4,5-Trihydroxy-2-pentanone, 1-Deoxy-D-threo-2-pentulose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10O4
CAS Number:
Molecular Weight:
134.13
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

Assay

≥80% (TLC)

form

liquid

optical activity

[α]/D +31.0±3.0°, c = 1 in H2O

storage temp.

−20°C

SMILES string

CC(=O)[C@@H](O)[C@H](O)CO

InChI

1S/C5H10O4/c1-3(7)5(9)4(8)2-6/h4-6,8-9H,2H2,1H3/t4-,5-/m1/s1

InChI key

IGUZJYCAXLYZEE-RFZPGFLSSA-N

Biochem/physiol Actions

Metabolite of the non-mevalonate pathway, generally found in prokaryotes, as precursor to isoprenoids as well as non-isoprenoids like vitamins. As this pathway is not present in humans, it is of interest for the development of bacterium-specific drugs in the search for treatments of infectious diseases.

Linkage

In E. coli, 1-deoxy-D-xylulose is converted into 1-deoxy-D-xylulose 5-phosphate by phosphorylation of the C-5 hydroxy group by D-xylulokinase.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andréa Hemmerlin et al.
The Journal of biological chemistry, 278(29), 26666-26676 (2003-05-09)
In plants, two pathways are utilized for the synthesis of isopentenyl diphosphate, the universal precursor for isoprenoid biosynthesis. The key enzyme of the cytoplasmic mevalonic acid (MVA) pathway is 3-hydroxy-3-methylglutaryl-coenzyme A reductase (HMGR). Treatment of Tobacco Bright Yellow-2 (TBY-2) cells
Ines Neundorf et al.
Chembiochem : a European journal of chemical biology, 4(11), 1201-1205 (2003-11-13)
Upon feeding of [2-(13)C,4-(2)H]-1-deoxy-D-xylulose to Streptomyces ghanaensis, the deuterium label was retained exclusively at positions C-7 and C-17 in the moenocinol part of the moenomycin antibiotics. This result vindicates the hypothesis that the C(25) structure of moenocinol is assembled from
Phosphorylation of 1-deoxy-D-xylulose by D-xylulokinase of Escherichia coli.
Wungsintaweekul, J.
European Journal of Biochemistry, 268, 310?316-310?316 (2001)
J Schwender et al.
Planta, 212(3), 416-423 (2001-04-06)
The biosynthesis of the C5 building block of isoprenoids, isopentenyl diphosphate (IPP), proceeds in higher plants via two basically different pathways; in the cytosolic compartment sterols are formed via mevalonate (MVA), whereas in the plastids the isoprenoids are formed via
Jiaqi Liu et al.
Frontiers in plant science, 8, 2082-2082 (2017-12-23)
As one type of the most important alkaloids in the world, terpenoid indole alkaloids (TIAs) show a wide range of pharmaceutical activities that are beneficial for clinical treatments.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service