Skip to Content
Merck
All Photos(1)

Key Documents

137254

SAFC

L-Tyrosine disodium salt dihydrate

EMPROVE® EVOLVE

Pharma Manufacturing

Synonym(s):

L-Tyrosine, Disodium Salt Dihydrate

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€242.00
1 KG
€916.00
10 KG
€5,960.00

€242.00


Please contact Customer Service for Availability


Select a Size

Change View
100 G
€242.00
1 KG
€916.00
10 KG
€5,960.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352209

€242.00


Please contact Customer Service for Availability

biological source

synthetic

Quality Level

product line

EMPROVE® EVOLVE

form

powder

technique(s)

cell culture | mammalian: suitable

impurities

Endotoxin, microbial, and heavy metals, tested

suitability

suitable for manufacturing use

foreign activity

Cytotoxicity, DNase, NICKase, RNase, and Protease; tested

storage temp.

2-30°C

SMILES string

[Na+].[Na+].N[C@@H](Cc1ccc(cc1)[O-])C(=O)[O-].O.O

InChI

1S/C9H11NO3.2Na.2H2O/c10-8(9(12)13)5-6-1-3-7(11)4-2-6;;;;/h1-4,8,11H,5,10H2,(H,12,13);;;2*1H2/q;2*+1;;/p-2/t8-;;;;/m0..../s1

InChI key

WFTCFVUQORELJZ-USHJOAKVSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Our SAFC® portfolio of high-quality raw materials for use in biopharmaceutical processing withstands strict quality control procedures plus the documentation and expertise to help our customers meet requirements as defined by the M-Clarity Program.


https://www.sigmaaldrich.com/US/en/life-science/quality-and-regulatory-management/m-clarity-program


As part of our EMPROVE® Program, our raw materials are offered with EMPROVE® Dossiers which provide comprehensive, up-to-date documentation to help you navigate regulatory challenges, manage risks, and improve your manufacturing processes. Our comprehensive portfolio of upstream process chemicals not only provides biopharmaceutical manufacturers with high-quality raw materials for production of classical and novel therapies, but also helps them get to market faster and simplify regulatory challenges. Trust us to deliver supply chain transparency and reliable sourcing around the globe, streamlining your product qualification with best-in-class regulatory support and service.

Application

L-Tyrosine disodium salt dihydrate is a sodium salt form of the amino acid L-Tyrosine, which is a polar, naturally occurring, non-essential amino acid. This compound can be utilized as a component in cell culture media for the production of therapeutic proteins, antibodies, vaccines, etc. L-Tyrosine disodium salt dihydrate generally exhibits greater solubility compared to L-Tyrosine

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service