Skip to Content
Merck
All Photos(1)

Key Documents

10419

Sigma-Aldrich

8-Anilino-1-naphthalenesulfonic acid hemimagnesium salt hydrate

for fluorescence, ≥95.0% (T)

Synonym(s):

1,8-ANS magnesium salt, ANSA magnesium salt, Magnesium 8-anilino-1-naphthalenesulfonate hydrate, N-Phenyl peri acid magnesium salt

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€36.10
25 G
€300.00

€36.10


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€36.10
25 G
€300.00

About This Item

Empirical Formula (Hill Notation):
C16H12Mg0.5NO3S · xH2O
CAS Number:
Molecular Weight:
310.49 (anhydrous basis)
Beilstein:
4834623
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

€36.10


Please contact Customer Service for Availability

Request a Bulk Order

grade

for fluorescence

Quality Level

Assay

≥95.0% (T)

form

powder

technique(s)

titration: suitable

solubility

hot water: 50 mg/mL, green

λmax

465-475 nm

fluorescence

λex 388 nm; λem 470 nm in 0.1 M Tris, 0.2 M KCl, pH 9.0, BSA

suitability

suitable for fluorescence

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

O.[Mg++].[O-]S(=O)(=O)c1cccc2cccc(Nc3ccccc3)c12.[O-]S(=O)(=O)c4cccc5cccc(Nc6ccccc6)c45

InChI

1S/2C16H13NO3S.Mg.H2O/c2*18-21(19,20)15-11-5-7-12-6-4-10-14(16(12)15)17-13-8-2-1-3-9-13;;/h2*1-11,17H,(H,18,19,20);;1H2/q;;+2;/p-2

InChI key

FBANPIUXAXTAFX-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

ANSA (8-Anilino-1-naphthalenesulfonic acid) is a hydrophobic polarity sensitive fluorescent dye useful as a site probe to detect conformational changes in cell and micelle membranes and molecules such as proteins. ANSA is also used as a fluorescent probe for the estimation of the critical micelle concentration (CMC) of surfactants.

Features and Benefits

Fluorescent probe for the estimation of the CMC of surfactants.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Bertsch et al.
Analytical biochemistry, 313(2), 187-195 (2003-02-28)
Hydrophobic sites on the surface of protein molecules are thought to have important functional roles. The identification of such sites can provide information about the function and mode of interaction with other cellular components. While the fluorescence enhancement of polarity-sensitive
J L Jensen et al.
Cellular and molecular biology (Noisy-le-Grand, France), 46(3), 685-696 (2000-06-29)
We have studied the metal-catalyzed oxidation (MCO) of brain-derived neurotrophic factor (BDNF) with regard to target sites and potential conformational changes of the protein. The exposure of BDNF to three different levels of ascorbate/Cu(II)/O2 [20 microM Cu(II), 2 mM ascorbate
C D Peters et al.
Biochemistry and cell biology = Biochimie et biologie cellulaire, 75(1), 55-61 (1997-01-01)
Protein aggregation is believed to be due to conformers that expose hydrophobic clusters that promote protein association. Such conformers can be detected using a fluorescent probe like 8-anilino 1-naphthalenesulfonic acid (ANSA). Here we show that urease exposed to 1.0 M
Rapid fluorescent "staining" of nondenatured protein bands in agar and polyacrylamide gels.
S T Nerenberg et al.
Analytical biochemistry, 43(2), 564-574 (1971-10-01)
Anilinonaphthalene sulfonate as a probe of membrane composition and function.
J Slavík
Biochimica et biophysica acta, 694(1), 1-25 (1982-08-11)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service