Skip to Content
Merck
All Photos(1)

Key Documents

08247

Sigma-Aldrich

3,4-Dimethyl-3-cyclohexene-1-carboxaldehyde

≥97.0% (GC)

Synonym(s):

3,4-Dimethyl-3- cyclohexenylmethanal, 3,4-Dimethyl-3-cyclohexenecarbaldehyde, 3,4-Dimethylcyclohex-3-enecarbaldehyde, 4-Formyl-1,2-dimethyl-1-cyclohexene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H14O
CAS Number:
Molecular Weight:
138.21
Beilstein:
2040252
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.32

Assay

≥97.0% (GC)

form

liquid

refractive index

n20/D 1.477-1.481

storage temp.

2-8°C

SMILES string

[H]C(C1CC(C)=C(C)CC1)=O

InChI

1S/C9H14O/c1-7-3-4-9(6-10)5-8(7)2/h6,9H,3-5H2,1-2H3

InChI key

NOJNKSAQZMVMFF-UHFFFAOYSA-N

Biochem/physiol Actions

Metabolite produced by plants, as a component of aroma fragrances.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Miguel Arruda et al.
Molecules (Basel, Switzerland), 17(3), 3082-3092 (2012-03-14)
Acetylcholinesterase inhibition, antioxidant and cytotoxic activities of Hedychium gardnerianum leaf essential oils from S. Miguel Island were determined. All the oils inhibited acetylcholinesterase, with IC(50) values of approximately 1 mg/mL, showing no statistical differences between collection sites. Three oils presented
Z L Cardeal et al.
Rapid communications in mass spectrometry : RCM, 20(19), 2823-2836 (2006-08-31)
The headspace compositions of 13 pepper and peppercorn samples of different species, colloquially also referred to as pepper, were analyzed, and more than 300 compounds were tentatively characterized by means of comprehensive two-dimensional gas chromatography in tandem with flame ionization
Chemical composition and antioxidant activity of the crude methanolic extracts of Mentha spicata.
Rameshwar Naidu, J., et al.
Journal of Phytology, 4, 13-18 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service