Skip to Content
Merck
All Photos(1)

Key Documents

07153

Sigma-Aldrich

7-Diethylamino-3-[N-(3-maleimidopropyl)carbamoyl]coumarin

BioReagent, suitable for fluorescence, ≥97.0% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H23N3O5
CAS Number:
Molecular Weight:
397.42
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.32

product line

BioReagent

Assay

≥97.0% (HPLC)

suitability

suitable for fluorescence

storage temp.

2-8°C

SMILES string

CCN(CC)c1ccc2C=C(C(=O)NCCCN3C(=O)C=CC3=O)C(=O)Oc2c1

InChI

1S/C21H23N3O5/c1-3-23(4-2)15-7-6-14-12-16(21(28)29-17(14)13-15)20(27)22-10-5-11-24-18(25)8-9-19(24)26/h6-9,12-13H,3-5,10-11H2,1-2H3,(H,22,27)

InChI key

PHFIKYNYJRZPND-UHFFFAOYSA-N

Application

Thiol-reactive probe for protein labelling[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thiol-reactive fluorescent probes for protein labelling
Corrie, J.E.T.
Journal of the Chemical Society. Perkin Transactions 1, 20, 2975-2978 (1994)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service