Skip to Content
Merck
All Photos(1)

Key Documents

06880

Sigma-Aldrich

4-Aminobenzamidine dihydrochloride

BioReagent, suitable for fluorescence, ≥99.0% (TLC)

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€222.00

€222.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€222.00

About This Item

Linear Formula:
H2NC6H4C(=NH)NH2·2HCl
CAS Number:
Molecular Weight:
208.09
Beilstein:
3692927
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.32

€222.00


Please contact Customer Service for Availability

Request a Bulk Order

product line

BioReagent

Assay

≥99.0% (TLC)

form

powder or crystals

mp

>300 °C (lit.)

fluorescence

λex >300 nm in H2O

suitability

suitable for fluorescence

storage temp.

2-8°C

SMILES string

Cl[H].Cl[H].NC(=N)c1ccc(N)cc1
Cl[H].Cl[H].NC(=N)c1ccc(N)cc1

InChI

1S/C7H9N3.2ClH/c8-6-3-1-5(2-4-6)7(9)10;;/h1-4H,8H2,(H3,9,10);2*1H

InChI key

GHEHNICLPWTXJC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Emmax: none

Other Notes

Competitive inhibitor of serine proteases[1][2]; As a fluorescent probe for the active site of serine proteases[3]

replaced by

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Comparative studies on the inhibition of trypsin, plasmin, and thrombin by derivatives of benzylamine and benzamidine.
F Markwardt et al.
European journal of biochemistry, 6(4), 502-506 (1968-12-05)
STUDIES ON THE ACTIVE CENTER OF TRYPSIN. THE BINDING OF AMIDINES AND GUANIDINES AS MODELS OF THE SUBSTRATE SIDE CHAIN.
M MARES-GUIA et al.
The Journal of biological chemistry, 240, 1579-1585 (1965-04-01)
S A Evans et al.
The Journal of biological chemistry, 257(6), 3014-3017 (1982-03-25)
p-Aminobenzamidine is weakly fluorescent in neutral aqueous buffer, with excitation and emission maxima at 293 and 376 nm, respectively. Binding to trypsin results in a blue shift of the emission peak to 362 nm, and 50-fold fluorescence enhancement, while binding
Nathan J Alves et al.
Biochemical and biophysical research communications, 457(3), 358-362 (2015-01-13)
The potent fibrinolytic enzyme, plasmin has numerous clinical applications for recannulizing vessels obstructed by thrombus. Despite its diminutive size, 91 kDa, success in the recombinant expression of this serine protease has been limited. For this reason, a truncated non-glycosylated plasmin

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service