Skip to Content
Merck
All Photos(1)

Key Documents

03181

Sigma-Aldrich

D-myo-Inositol 1,2,3,5,6-pentakis-phosphate

≥80% (HPLC)

Synonym(s):

L-myo-Inositol 1,2,3,4,5-pentakis-phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H17O21P5
CAS Number:
Molecular Weight:
580.06
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.32
Pricing and availability is not currently available.

Quality Level

Assay

≥80% (HPLC)

storage temp.

−20°C

SMILES string

O[C@@H]1[C@@H](OP(O)(O)=O)[C@@H](OP(O)(O)=O)[C@@H](OP(O)(O)=O)[C@H](OP(O)(O)=O)[C@H]1OP(O)(O)=O

InChI

1S/C6H17O21P5/c7-1-2(23-28(8,9)10)4(25-30(14,15)16)6(27-32(20,21)22)5(26-31(17,18)19)3(1)24-29(11,12)13/h1-7H,(H2,8,9,10)(H2,11,12,13)(H2,14,15,16)(H2,17,18,19)(H2,20,21,22)/t1-,2-,3+,4-,5-,6-/m1/s1

InChI key

CTPQAXVNYGZUAJ-UOTPTPDRSA-N

General description

Aluminum was shown to increase the activity of multiple inositol polyphosphate phosphatathase(MIPP) in hepatic mammalian cells. This cased a decrease in the MIPP substrate, Ins(1,2,3,5,6)P5. [1]

Biochem/physiol Actions

The metabolite inositol is a precursor of inositides, which have an important impact on diverse areas of cellular regulation[2].

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Ali et al.
The Biochemical journal, 305 ( Pt 2), 557-561 (1995-01-15)
There is speculation that some of the toxic effects of Al3+ may originate from it perturbing inositol phosphate/Ca2+ signalling. For example, in permeabilized L1210 mouse lymphoma cells, 10-50 microM Al3+ activated Ins(1,3,4,5)P4-dependent Ca2+ mobilization and Ins(1,3,4,5)P4 3-phosphatase activity [Loomis-Husselbee, Cullen
Christopher J Barker et al.
Methods in molecular biology (Clifton, N.J.), 645, 1-19 (2010-07-21)
Inositides have an important impact on diverse areas of cellular regulation. However, since this area has grown exponentially from the mid 1980s onwards, many workers find themselves relatively new to the field. In this chapter, we establish a broad foundation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service