Skip to Content
Merck
All Photos(1)

Key Documents

01230

Sigma-Aldrich

Acetyl isothiocyanate

≥97.0% (GC)

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
€182.00

€182.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 ML
€182.00

About This Item

Linear Formula:
CH3CON=C=S
CAS Number:
Molecular Weight:
101.13
Beilstein:
635801
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.26

€182.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥97.0% (GC)

form

liquid

refractive index

n20/D 1.532

bp

132-133 °C (lit.)

density

1.151 g/mL at 20 °C (lit.)

storage temp.

−20°C

SMILES string

CC(=O)N=C=S
CC(=O)N=C=S

InChI

1S/C3H3NOS/c1-3(5)4-2-6/h1H3

InChI key

VITFJKNVGRZRKB-UHFFFAOYSA-N

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eva A M Schaefer et al.
Biochemical pharmacology, 85(3), 426-438 (2012-12-12)
TRPA1, a member of the transient receptor potential (TRP) family of cation channels, has mainly been characterized as a chemosensory protein in neuronal cells. TRPA1 is activated by toxic or irritating volatile agents like allyl isothiocyanate (AITC), tear gas, formalin
Stephen Bell
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 61(7), 1471-1477 (2005-04-12)
A wide range of ab initio and hybrid density functional methods and basis sets have been employed to calculate the barriers to methyl internal rotation in a range of molecules with the acetyl moiety. Comparison is made of the computed
B Mo et al.
Analytical biochemistry, 252(1), 169-176 (1997-11-05)
A new derivatizing reagent, acetyl isothiocyanate (AITC), is applied for C-terminal peptide sequencing. It has been successfully used to sequence six C-terminal residues of a synthetic peptide at low nanomole levels. According to the mechanism study of the derivatization of

Questions

  1. What is the colour of acetyl isothiocyanate

    1 answer
    1. This product has a color range from yellow to very dark yellow to very dark orange. The exact color will be provided on the lot specific certificate of analysis.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service