Skip to Content
Merck
All Photos(8)

Key Documents

793523

Sigma-Aldrich

Zinc chloride

Zinc chloride

anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%

Synonym(s):

Dichlorozinc

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€27.50
500 G
€93.70
1 KG
€158.00

€27.50


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 G
€27.50
500 G
€93.70
1 KG
€158.00

About This Item

Linear Formula:
ZnCl2
CAS Number:
Molecular Weight:
136.30
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
grade:
anhydrous
reagent grade
form:
powder or crystals

€27.50


Please contact Customer Service for Availability

Request a Bulk Order

grade

anhydrous
reagent grade

Quality Level

vapor pressure

1 mmHg ( 428 °C)

product line

Redi-Dri

Assay

≥98%

form

powder or crystals

quality

free-flowing

pH

5 (20 °C, 100 g/L)

mp

293 °C (lit.)

SMILES string

[Cl-].[Cl-].[Zn++]

InChI

1S/2ClH.Zn/h2*1H;/q;;+2/p-2

InChI key

JIAARYAFYJHUJI-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Zinc chloride is a Lewis acid that can be used to promote cycloaddition, substitution, and addition reactions, including electrophilic aromatic additions and selective reductions. It is also used in the preparation of organozinc reagents via transmetalation.

Application

Zinc chloride can be used as a catalyst in the:
  • Chemoselective dimethyl acetalization of aldehydes and ketones with trimethyl orthoformate to synthesize dimethyl acetals.
  • Synthesis of 9-substituted acridines via Bernthsen reaction.
It can also be used as an additive in the preparation of ultrafiltration membranes.

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elisabeth Elje et al.
Nanomaterials (Basel, Switzerland), 10(3) (2020-03-22)
(1) In compliance with the 3Rs policy to reduce, refine and replace animal experiments, the development of advanced in vitro models is needed for nanotoxicity assessment. Cells cultivated in 3D resemble organ structures better than 2D cultures. This study aims
Shunfeng Wang et al.
Molecules (Basel, Switzerland), 25(21) (2020-11-14)
In the present study, the fabrication of a biomimetic wound dressing that mimics the extracellular matrix, consisting of a hydrogel matrix composed of non-oxidized and periodate-oxidized marine alginate, was prepared to which gelatin was bound via Schiff base formation. Into

Articles

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

Active Filters

  1. Oregon
    • Review 1
    • Votes 0
    5 out of 5 stars.

    ZnCl2 for pollen flotation

    I typically use ZnBr2 to raise the raise the specific gravity for pollen suspension. However, I could not afford it. I opted to try with the ZnCl2, and it worked perfectly! The powder was super clean and floated the pollen out perfectly! I’m grateful to M-S that they had material I could afford of such high quality!!

    Helpful?

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service