Skip to Content
Merck
All Photos(1)

Key Documents

673781

Sigma-Aldrich

Acetone

ACS reagent, ≥99.5%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH3
CAS Number:
Molecular Weight:
58.08
Beilstein:
635680
EC Number:
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent

Quality Level

vapor density

2 (vs air)

vapor pressure

184 mmHg ( 20 °C)

Assay

≥99.5%

form

liquid

expl. lim.

13.2 %

impurities

≤0.0003 meq/g Titr. acid
≤0.0006 meq/g Titr. base
≤0.002% aldehyde as formaldehyde
≤0.05% isopropanol
≤0.05% methanol
≤0.5% water

evapn. residue

≤0.001%

color

APHA: ≤10

refractive index

n20/D 1.359 (lit.)

pH

5-6 (20 °C, 395 g/L)

bp

56 °C/760 mmHg (lit.)

mp

−94 °C (lit.)

solubility

water: miscible

density

0.791 g/mL at 25 °C (lit.)

format

neat

SMILES string

CC(C)=O

InChI

1S/C3H6O/c1-3(2)4/h1-2H3

InChI key

CSCPPACGZOOCGX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Acetone is an organic solvent widely used in the polymer and pharmaceutical industry. It is found in mammalian tissues as a by-product of metalbolism.1 Its anticonvulsant property has been studied. Acetone in combination with tetraalkylammonium chloride forms an efficient solvent system for dissolving cellulose. P84 (polyimide membrane)/TAEA (tripodal amine (tris(2-aminoethyl)amine) cross-linked asymmetric flat sheet membranes based pervaporation for the dehydration of acetone has been reported. A recent study reports the photolysis of acetone vapors in a titanium dioxide (TiO2) photocatalyst immobilized, UV irradiated reactor.

Application

Acetone may be employed in the following studies:
  • As a precursor for the synthesis of methyl isobutyl ketone (MIBK) in the presence of sulfonated graphene oxide-Pd/cordierite catalyst.
  • Synthesis of (4-hydroxymethyl-2,2-dimethyl-1,3-dioxolane), a solketal from glycerol using supercritical fluids (SCF) technology.
  • As a starting material to synthesize methyl methacrylate.1
It may be used in the synthesis of the following acetone hydrazones:
  • 1-isopropylidene-2-methylhydrazine
  • 1-isopropylidene-2-hydroxyethylhydrazine
  • 1-isopropylidene-2-formylhydrazine
Acetone′s luminesence intensity is dependent upon the solution components . The absorption of UV light by acetone, results in its photolysis and the production of radials .

greener alternative product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Acetone as an anticonvulsant.
Likhodii S, et al.
Epilepsia, 49(8), 83-86 (2008)
Marc Kostag et al.
Macromolecular rapid communications, 35(16), 1419-1422 (2014-06-14)
Acetone containing tetraalkylammonium chloride is found to be an efficient solvent for cellulose. The addition of an amount of 10 mol% (based on acetone) of well-soluble salt triethyloctylammonium chloride (Et3 OctN Cl) adjusts the solvent's properties (increases the polarity) to
Sulfonated graphene oxide supported Pd bifunctional catalyst for one-pot synthesis of methyl isobutyl ketone from acetone with high conversion and selectivity.
Liu M, et al.
J. Mol. Catal. A: Chem., 408, 85-90 (2015)
P84 polyimide membranes modified by a tripodal amine for enhanced pervaporation dehydration of acetone.
Mangindaan DW, et al.
Chemical Engineering Science, 122, 14-23 (2015)
Ketalization of glycerol to solketal in supercritical acetone.
Royon D, et al.
Journal of Supercritical Fluids, 58(1), 88-92 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service