Skip to Content
Merck
All Photos(1)

Key Documents

576905

Sigma-Aldrich

Water solution

contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC

Synonym(s):

Aqueous Solution Standard

Sign Into View Organizational & Contract Pricing

Select a Size

18 L
€595.00
20 L
€907.00

€595.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
18 L
€595.00
20 L
€907.00

About This Item

Linear Formula:
H2O
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:
NACRES:
NA.21
grade:
HPLC grade
technique(s):
HPLC: suitable
application(s):
food and beverages

€595.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

HPLC grade

form

liquid

contains

0.1 % (v/v) trifluoroacetic acid

technique(s)

HPLC: suitable

UV absorption

λ: 210 nm Amax: ≤0.60
λ: 230 nm Amax: <0.07
λ: 254 nm Amax: ≤0.005

application(s)

food and beverages

SMILES string

O

InChI

1S/H2O/h1H2

InChI key

XLYOFNOQVPJJNP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The product is water containing 0.1%v/v trifluoroacetic acid. This pre-blended mobile phase solvent is of high purity that reduces time-consuming preparation and prevents instrument downtime because of impure mobile phases. It can be employed for HPLC and UHPLC (Ultra High performance Liquid Chromatography) techniques. Trifluoroacetic acid (TFA) is commonly used as a mobile phase additive. It is a UV transparent, volatile acid that forms hydrophobic negatively charged trifluoroacetate ion.[1]

Preparation Note

Product filtered through a 0.2 μm filter

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Optimum concentration of trifluoroacetic acid for reversed-phase liquid chromatography of peptides revisited.
Chen Y, et al.
Journal of Chromatography A, 1043(1), 9-18 (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service