Skip to Content
Merck
All Photos(1)

Key Documents

438065

Sigma-Aldrich

Hydrobromic acid

ACS reagent, 48%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HBr
CAS Number:
Molecular Weight:
80.91
Beilstein:
3587158
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

grade

ACS reagent

vapor density

2.8 (vs air)

vapor pressure

320 psi ( 21.1 °C)
8 mmHg ( 25 °C)

description

48% aqueous HBr

Assay

48%

form

liquid

ign. residue

≤0.002%

density

1.49 g/mL at 25 °C (lit.)

anion traces

SO42- and SO32-: ≤0.003%
chloride (Cl-): ≤0.05%
iodide (I-): ≤0.003%
phosphate (PO43-): ≤0.001%

cation traces

Fe: ≤1 ppm
Se: ≤0.01 ppm
heavy metals (as Pb): ≤5 ppm

SMILES string

Br

InChI

1S/BrH/h1H

InChI key

CPELXLSAUQHCOX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ahmed Hasbi et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 28(11), 4806-4820 (2014-07-27)
Although the dopamine D1-D2 receptor heteromer has emerging physiological relevance and a postulated role in different neuropsychiatric disorders, such as drug addiction, depression, and schizophrenia, there is a need for pharmacological tools that selectively target such receptor complexes in order
Yaohui Tang et al.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism, 34(7), 1138-1147 (2014-04-10)
Neural stem cells (NSCs) show therapeutic potential for ischemia in young-adult animals. However, the effect of aging on NSC therapy is largely unknown. In this work, NSCs were transplanted into aged (24-month-old) and young-adult (3-month-old) rats at 1 day after
Po-Yu Tsai et al.
Physical chemistry chemical physics : PCCP, 13(4), 1419-1423 (2010-11-27)
The orientation dependence of Br-atom formation in the reaction of the oriented OH radical with the HBr molecule using the hexapole electrostatic field was studied. Experimental results for the orientation dependence in the reaction were analyzed using a Legendre polynomial
Małgorzata Olejniczak et al.
The Journal of chemical physics, 136(1), 014108-014108 (2012-01-14)
We report the implementation of nuclear magnetic resonance (NMR) shielding tensors within the four-component relativistic Kohn-Sham density functional theory including non-collinear spin magnetization and employing London atomic orbitals to ensure gauge origin independent results, together with a new and efficient
Peter Comba et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 18(6), 1700-1710 (2012-01-12)
The synthesis and characterization of a novel dinucleating ligand L (L=4,11-dimethyl-1,8-bis{2-[N-(di-2-pyridylmethyl)amino]ethyl}cyclam) and its μ-oxo-bridged diferric complex [(H(2)L){Fe(III)(2)(O)}(Cl)(4)](2+) are reported. This diiron(III) complex is the first example of a truly functional purple acid phosphatase (PAP) mimic as it accelerates the hydrolysis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service