Skip to Content
Merck
All Photos(2)

Key Documents

345695

Sigma-Aldrich

Titanium(IV) chloride solution

1.0 M in toluene

Synonym(s):

Titanium tetrachloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
TiCl4
CAS Number:
Molecular Weight:
189.68
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.55

form

liquid

reaction suitability

reagent type: catalyst
core: titanium

concentration

0.95-1.10 M NaOH (titration)
1.0 M in toluene

density

0.964 g/mL at 25 °C

SMILES string

Cl[Ti](Cl)(Cl)Cl

InChI

1S/4ClH.Ti/h4*1H;/q;;;;+4/p-4

InChI key

XJDNKRIXUMDJCW-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

General description

Titanium(IV) chloride participates in the Baylis-Hillman reaction of arylaldehydes with methyl vinyl ketone.

Application

TiCl4 can be used as a precursor for the synthesis of nanosized titaniuim dioxide.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Aquatic Chronic 3 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Repr. 2 - Skin Corr. 1B - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

46.4 °F - closed cup

Flash Point(C)

8 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Titanium(IV) chloride and the amine-promoted baylis-hillman reaction
Shi et al.
Organic letters, 2(23), 3755-3755 (2000-11-14)
Precipitation of nanosized titanium dioxide from aqueous titanium (IV) chloride solutions by neutralization with MgO.
Li Y and Demopoulos GP.
Hydrometallurgy, 90(1), 26-33 (2008)
Yang Chen et al.
ChemSusChem, 4(10), 1498-1503 (2011-09-29)
Mesoporous TiO₂ beads with diameters of 320±50, 550±50, and 830±40 nm, comprising interconnected and densely packed TiO₂ nanocrystals, were used as working electrodes for dye-sensitized solar cells (DSCs). These electrodes possess high surface areas and superior light-scattering properties, which are ideal
Bratindranath Mukherjee et al.
Nanoscale, 5(1), 269-274 (2012-11-20)
The surface treatment of an anodized TiO(2) nanotube (T_NT) is very desirable for enhancing its photoelectrochemical properties and often is a prerequisite to deposition of any overlying layer for photoactivity efficiency improvement. This study provides a comparative analysis of the
Kazuhiko Akiyama et al.
Journal of the American Chemical Society, 134(23), 9762-9767 (2012-05-18)
Rapid and efficient separation/purification of pure metallofullerenes M(x)@C(n) (M = metal; x = 1, 2; n > 70) and carbide metallofullerenes of the type M(y)C(2)@C(n-2) (y = 2, 3, 4; n - 2 > 68) has been reported. The present

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service