Skip to Content
Merck
All Photos(1)

Key Documents

30107

Sigma-Aldrich

Hydrofluoric acid

puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥48%

Synonym(s):

HF

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HF
CAS Number:
Molecular Weight:
20.01
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.21
Agency:
reag. ISO
reag. Ph. Eur.

grade

ACS reagent
puriss. p.a.

Quality Level

Agency

reag. ISO
reag. Ph. Eur.

vapor density

1.27 (vs air)

vapor pressure

25 mmHg ( 20 °C)

Assay

≥48%

form

liquid

impurities

≤0.005% hexafluorosilic acid (H2SiF6)

ign. residue

≤0.0005% (as SO4)

density

1.15 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤1 mg/kg
phosphate (PO43-): ≤0.5 mg/kg
sulfate (SO42-): ≤2 mg/kg
sulfite (SO32-): ≤2 mg/kg

cation traces

Ag: ≤0.02 mg/kg
Al: ≤0.05 mg/kg
As: ≤0.05 mg/kg
Ba: ≤0.1 mg/kg
Be: ≤0.02 mg/kg
Bi: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.01 mg/kg
Co: ≤0.02 mg/kg
Cr: ≤0.02 mg/kg
Cu: ≤0.02 mg/kg
Fe: ≤0.2 mg/kg
Ge: ≤0.05 mg/kg
K: ≤0.1 mg/kg
Li: ≤0.02 mg/kg
Mg: ≤0.2 mg/kg
Mn: ≤0.05 mg/kg
Mo: ≤0.05 mg/kg
Na: ≤0.2 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.05 mg/kg
Sr: ≤0.02 mg/kg
Ti: ≤0.1 mg/kg
Tl: ≤0.05 mg/kg
V: ≤0.05 mg/kg
Zn: ≤0.05 mg/kg
Zr: ≤0.1 mg/kg

SMILES string

[H+].[F-]

InChI

1S/FH/h1H

InChI key

KRHYYFGTRYWZRS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hydrofluoric acid (HF) is a strong inorganic acid that is commonly used as a source of fluorine for the production of organofluorine compounds and inorganic fluorides. It is also used as a deprotecting reagent in organic synthesis. Additionally, it is also used for breaking strong Si-O bonds to form SiF6− 2 ions in an acidic solution.

Application


  • Speciation of tin ions in oxide glass containing iron oxide through solvent extraction and inductively coupled plasma atomic emission spectrometry after the decomposition utilizing ascorbic acid.: This study explores the speciation of tin ions in iron oxide-containing glass, utilizing ascorbic acid for decomposition followed by solvent extraction and analysis via inductively coupled plasma atomic emission spectrometry, highlighting a novel analytical chemistry application of hydrofluoric acid (Saijo Y et al., 2022).

  • Physicochemical and morphological characterization of a glass ceramic treated with different ceramic primers and post-silanization protocols.: This paper discusses the effects of various ceramic primers and silanization protocols on the physicochemical properties and morphology of glass ceramics, contributing to analytical chemistry by elucidating the interactions and stability of treated surfaces (Moreno MBP et al., 2019).

  • Effect of Surface Cleaning Regimen on Glass Ceramic Bond Strength.: Investigates the impact of different surface cleaning regimens, including the use of hydrofluoric acid, on the bond strength of glass ceramics, providing insights into optimal surface treatment methods in material science and analytical chemistry (Lapinska B et al., 2019).

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Highly-ordered TiO2 nanotube arrays up to 220μm in length: use in water photoelectrolysis and dye-sensitized solar cells.
Shankar K, et al.
Nanotechnology, 18(6), 065707-065707 (2007)
Anatase TiO2 nanosheet: an ideal host structure for fast and efficient lithium insertion/extraction.
Chen JS and Lou XW.
Electrochemical Communications, 11(12), 2332-2335 (2009)
Eagleson M.
Concise Encyclopedia Chemistry, 508-508 (1994)
Francesca Monticelli et al.
Dental materials : official publication of the Academy of Dental Materials, 22(7), 602-609 (2005-11-18)
To verify the influence of different etching procedures of the post-surface on microtensile bond strength values between fiber posts and composite core materials. 60 DT Light Posts were divided into 10 subgroups using five different chemical surface treatments and two
Nicolas Vodovar et al.
JACC. Heart failure, 3(8), 629-636 (2015-08-08)
This study sought to hypothesize that elevated B-type natriuretic peptide (BNP) could act as an endogenous neprilysin inhibitor. A hallmark of acute decompensated heart failure (ADHF) is the overproduction of natriuretic peptides (NPs) by stretched cardiomyocytes. Various strategies have been

Articles

Professor Gogotsi and Dr. Shuck introduce MXenes: a promising family of two-dimensional materials with a unique combination of high conductivity, hydrophilicity, and extensive tunability.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service