Skip to Content
Merck
All Photos(2)

Key Documents

259705

Sigma-Aldrich

Methyl formate

spectrophotometric grade, ≥98%

Synonym(s):

Formic acid methyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
€184.00

€184.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 L
€184.00

About This Item

Linear Formula:
HCO2CH3
CAS Number:
Molecular Weight:
60.05
Beilstein:
1734623
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.21
grade:
spectrophotometric grade
technique(s):
UV/Vis spectroscopy: suitable
bp:
32-34 °C (lit.)
vapor pressure:
32.81 psi ( 55 °C)
9.21 psi ( 20 °C)

€184.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

spectrophotometric grade

Quality Level

vapor density

2.1 (vs air)

vapor pressure

32.81 psi ( 55 °C)
9.21 psi ( 20 °C)

Assay

≥98%

form

liquid

autoignition temp.

842 °F

expl. lim.

23 %

technique(s)

UV/Vis spectroscopy: suitable

impurities

<0.010% water

evapn. residue

<0.0005%

refractive index

n20/D 1.343 (lit.)

pH

4.0-5.0 (20 °C, 200 g/L)

bp

32-34 °C (lit.)

mp

−100 °C (lit.)

density

0.974 g/mL at 20 °C (lit.)

λ

H2O reference

UV absorption

λ: 259 nm Amax: 1.00
λ: 260 nm Amax: 0.70
λ: 265 nm Amax: 0.20
λ: 270 nm Amax: 0.04
λ: 310-400 nm Amax: 0.01

SMILES string

[H]C(=O)OC

InChI

1S/C2H4O2/c1-4-2-3/h2H,1H3

InChI key

TZIHFWKZFHZASV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl formate (MF) is one of the most significant intermediates in C1 chemistry. MF has been used to produce methanamide, N, N-dimethylformamide, dimethyl carbonate, methyl glycolate, acetic acid, ethylene glycol, methyl methacrylate, and high-purity CO. Additionally, MF can be used as a solvent for nitrocellulose and cellulose acetate.

Application


  • Bifunctionality of Re Supported on TiO(2) in Driving Methanol Formation in Low-Temperature CO(2) Hydrogenation.: This article explores the catalytic roles of methyl formate in methanol production from CO2 hydrogenation, providing a pathway for sustainable chemical synthesis (Urakawa A et al., 2023).

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Flam. Liq. 1 - STOT SE 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-2.2 °F - closed cup

Flash Point(C)

-19 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Methyl N-Phenyl Carbamate Synthesis from Aniline and Methyl Formate: Carbon Recycling to Chemical Products.
Yalfani MS, et al.
ChemSusChem, 8(3), 443-447 (2015)
Katherine R Phillips et al.
Journal of the American Chemical Society, 135(2), 574-577 (2012-12-28)
Methyl formate is produced from the photo-oxidation of methanol on preoxidized TiO(2)(110). We demonstrate that two consecutive photo-oxidation steps lead to methyl formate using mass spectrometry and scanning tunneling microscopy. The first step in methanol oxidation is formation of methoxy
Barriers to rotation adjacent to double bonds. 3. The carbon-oxygen barrier in formic acid, methyl formate, acetic acid, and methyl acetate. The origin of ester and amide resonance.
Wiberg KB and Laidig KE.
Journal of the American Chemical Society, 109(20), 5935-5943 (1987)
Methyl formate as a carbonylating agent for the catalytic conversion of phenol to methyl phenyl carbonate.
Wu C, et al.
Green Chemistry, 17(3), 1467-1472 (2015)
Microwave spectrum, barrier to internal rotation, and structure of methyl formate.
Curl Jr RF.
J. Chem. Phys. , 30(6), 1529-1536 (1959)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service