Skip to Content
Merck
All Photos(1)

Key Documents

236098

Sigma-Aldrich

Aluminum fluoride trihydrate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
AlF3·3H2O
CAS Number:
Molecular Weight:
138.02
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
grade:
for analytical purposes
form:
powder

grade

for analytical purposes

Assay

97%

form

powder

density

2.88 g/mL at 25 °C (lit.)

SMILES string

[H]O[H].[H]O[H].[H]O[H].F[Al](F)F

InChI

1S/Al.3FH.3H2O/h;3*1H;3*1H2/q+3;;;;;;/p-3

InChI key

BKLZIAYVINRQEJ-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Crystallographic Evidence for the Trihydrate of Aluminum Fluoride.
Freeman RD.
The Journal of Physical Chemistry, 61(2), 256-256 (1957)
The Thermal Decomposition of Ammonium Hexafluoroferrate (III) and Ammonium Hexafluoroaluminate. A New Crystalline Form of Aluminum Fluoride.
Shinn DB, et al.
Inorganic Chemistry, 5(11), 1927-1933 (1966)
Growth and dissolution kinetics of aluminium fluoride trihydrate crystals.
Nielsen AE and Altintas ND.
Journal of Crystal Growth, 69(2), 213-230 (1984)
Buchel KH, et al.
Industrial Inorganic Chemistry, 1-1 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service