Skip to Content
Merck
All Photos(2)

Key Documents

213357

Sigma-Aldrich

Mercury(II) oxide red

ACS reagent, ≥99.0%

Synonym(s):

Mercuric oxide red

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HgO
CAS Number:
Molecular Weight:
216.59
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NB.24

grade

ACS reagent

Assay

≥99.0%

form

powder

reaction suitability

reagent type: catalyst
core: mercury

impurities

≤0.005% N compounds
≤0.03% insol. dil. HCl

reduction residue

≤0.025%

mp

500 °C (dec.) (lit.)

anion traces

chloride (Cl-): ≤0.025%
sulfate (SO42-): ≤0.015%

cation traces

Fe: ≤0.005%

SMILES string

O=[Hg]

InChI

1S/Hg.O

InChI key

UKWHYYKOEPRTIC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Target Organs

Kidney

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Chambers C and Holliday AK.
Inorganic Chemistry, 381-381 (2013)
S R Krystek et al.
International journal of peptide and protein research, 34(1), 52-55 (1989-07-01)
Treatment of peptides with excess HgO in the presence of alkaline cyanide leads to cleavage of the peptides at glycine residues. The reaction appears to involve both C- and N-mercuration with subsequent release of 2 mol mercury per mol of
Bingyan Han et al.
Analytical chemistry, 81(13), 5569-5573 (2009-06-09)
Herein, a sensitive and selective sensor for biothiols based on the recovered fluorescence of the CdTe quantum dots (QDs)-Hg(II) system is reported. Fluorescence of QDs could be quenched greatly by Hg(II). In the presence of biothiols, such as glutathione (GSH)
B Kamoun et al.
Journal francais d'ophtalmologie, 22(5), 525-527 (1999-07-27)
Demodex folliculorum has been incriminated in the development of blepharitis although much controversy persists. Certain authors suggest that Demodex is a direct pathogen in chronic palpebral conditions while others consider the saprophyte to be innocuous to skin. We conducted a
R A Hyndiuk et al.
Antimicrobial agents and chemotherapy, 34(4), 610-613 (1990-04-01)
A double-masked, placebo-controlled, randomized study was done to assess the safety and clinical and quantitative microbiologic efficacy of 1% mercuric oxide (yellow) ophthalmic ointment in the treatment of eyelid infections, i.e., bacterial blepharitis. A total of 39 patients with bacterial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service