Key Documents
Z413887
Sigma-Aldrich
Aldrich® reducing adapter
female joint: ST/NS 24/40, male joint: ST/NS 34/45
Synonym(s):
glass adapter, glass adapters
Select a Size
1 EA
€86.30
€86.30
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
€86.30
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
€86.30
Please contact Customer Service for Availability
Joint
female joint: ST/NS24/40
male joint: ST/NS34/45
Related Categories
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)