Skip to Content
Merck
All Photos(1)

Documents

Y0001320

Ondansetron impurity F

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

2-Methylimidazole, 2-Methylglyoxaline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6N2
CAS Number:
Molecular Weight:
82.10
Beilstein:
1368
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

vapor pressure

<1 mmHg ( 0 °C)

API family

ondansetron

manufacturer/tradename

EDQM

bp

267-268 °C (lit.)

mp

142-143 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cc1ncc[nH]1

InChI

1S/C4H6N2/c1-4-5-2-3-6-4/h2-3H,1H3,(H,5,6)

InChI key

LXBGSDVWAMZHDD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Ondansetron impurity F EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Repr. 1B - Skin Corr. 1C

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mengzhe Guo et al.
Analytica chimica acta, 865, 31-38 (2015-03-04)
Matrix-assisted laser desorption ionization (MALDI) time-of-flight mass spectrometry (TOF-MS) can be considered as state of the art in the field of proteins and peptides analysis. In this work, we have designed an ionic liquid derivative strategy to obtain abundant fragment
Yonghai Song et al.
The Analyst, 140(2), 623-629 (2014-11-27)
A simple strategy to fabricate a fluorescent SiO2@zeolitic imidazolate framework-8 (ZIF-8) core-shell nanosensor for Cu(2+) detection was demonstrated in this work. The nanosensor was synthesized using carboxyl-functionalized SiO2 nanoparticles (SiO2 NPs) as a template to induce the growth of ZIF-8
Xiehong Cao et al.
Angewandte Chemie (International ed. in English), 53(5), 1404-1409 (2014-01-25)
A simple method for the preparation of metal-oxide-coated three-dimensional (3D) graphene composites was developed. The metal-organic frameworks (MOFs) that served as the precursors of the metal oxides were first synthesized on the 3D graphene networks (3DGNs). The desired metal oxide/3DGN
Hangzhen Lan et al.
Journal of chromatography. A, 1365, 35-44 (2014-09-15)
A high-performance magnetic molecularly imprinted polymer (MIP) coating using zeolite imidazolate framework-8 coated magnetic iron oxide (Fe3O4@ZIF-8) as a carrier was developed for simultaneous automated solid phase microextraction of four estrogens in 24 food samples. The coating material, abbreviated as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service