Skip to Content
Merck
All Photos(1)

Key Documents

Y0001238

Sulfinpyrazone for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(±)-Sulfinpyrazone, 1,2-Diphenyl-4-(phenylsulfinylethyl)-3,5-pyrazolidinedione, Diphenylpyrazone

Sign Into View Organizational & Contract Pricing

Select a Size

0.5 MG
€107.00

€107.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
0.5 MG
€107.00

About This Item

Empirical Formula (Hill Notation):
C23H20N2O3S
CAS Number:
Molecular Weight:
404.48
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€107.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

sulfinpyrazone

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O=C1C(CCS(=O)c2ccccc2)C(=O)N(N1c3ccccc3)c4ccccc4

InChI

1S/C23H20N2O3S/c26-22-21(16-17-29(28)20-14-8-3-9-15-20)23(27)25(19-12-6-2-7-13-19)24(22)18-10-4-1-5-11-18/h1-15,21H,16-17H2

InChI key

MBGGBVCUIVRRBF-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Sulfinpyrazone for system suitability EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E H Margulies et al.
Drugs, 20(3), 179-197 (1980-09-01)
Sulfinpyrazone1 has long been recognised as a potent uricosuric agent, but has more recently been studied extensively as a platelet inhibitor and antithrombotic agent. It is active in man following oral administration and has been reported to be effective in
Sulfinpyrazone: relationship between dose, kinetics, plasma concentrations and biological effects.
M R Buchanan
Thrombosis research. Supplement, 4, 89-92 (1983-01-01)
Uricosuric drugs, with special reference to probenecid and sulfinpyrazone.
A B Gutman
Advances in pharmacology, 4, 91-142 (1966-01-01)
Andrew C Kotze et al.
Antimicrobial agents and chemotherapy, 58(12), 7475-7483 (2014-10-08)
We used an enzyme induction approach to study the role of detoxification enzymes in the interaction of the anthelmintic compound naphthalophos with Haemonchus contortus larvae. Larvae were treated with the barbiturate phenobarbital, which is known to induce the activity of
Ashley M Laughney et al.
Science translational medicine, 6(261), 261ra152-261ra152 (2014-11-08)
Eribulin mesylate was developed as a potent microtubule-targeting cytotoxic agent to treat taxane-resistant cancers, but recent clinical trials have shown that it eventually fails in many patient subpopulations for unclear reasons. To investigate its resistance mechanisms, we developed a fluorescent

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service