Skip to Content
Merck
All Photos(1)

Key Documents

Y0000616

Tropisetron hydrochloride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Tropisetron monohydrochloride, 1H-Indole-3-carboxylic acid (3-endo)-8-methyl-8-azabicyclo[3.2.1]oct-3-yl ester monohydrochloride, 3-Tropanylindole-3-carboxylate monohydrochloride, ICS-205,930, Navoban

Sign Into View Organizational & Contract Pricing

Select a Size

40 MG
€132.00

€132.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
40 MG
€132.00

About This Item

Empirical Formula (Hill Notation):
C17H20N2O2 · HCl
CAS Number:
Molecular Weight:
320.81
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€132.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

tropisetron

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cl[H].CN1C2CCC1CC(C2)OC(=O)c3c[nH]c4ccccc34

InChI

1S/C17H20N2O2.ClH/c1-19-11-6-7-12(19)9-13(8-11)21-17(20)15-10-18-16-5-3-2-4-14(15)16;/h2-5,10-13,18H,6-9H2,1H3;1H

InChI key

XIEGSJAEZIGKSA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Tropisetron hydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Selective 5-HT3 serotonin receptor antagonist.
Tropisetron a selective serotonin 5-HT3 receptor antagonist is used mainly as an antiemetic to treat nausea and vomiting following chemotherapy . In the treatment of fibromyalgia syndrome (FMS) tropisetron reduced not only pain perception but also had a favourable effect on cardiac dysfunction during treatment.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Orie Tomoyose et al.
Journal of pharmacological sciences, 126(2), 136-145 (2014-09-26)
Fluvoxamine, a selective serotonin (5-HT) reuptake inhibitor, has been shown to exert analgesic effects in humans and laboratory animals. However, its effects on spinal nociceptive synaptic transmission have not been fully characterized. Here, whole-cell recordings were made from dorsal horn

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service