Y0000444
Flunixin impurity B
European Pharmacopoeia (EP) Reference Standard
Synonym(s):
2-Methyl-3-(trifluoromethyl)aniline, 2-Amino-6-(trifluoromethyl)toluene
About This Item
Recommended Products
grade
pharmaceutical primary standard
API family
flunixin
manufacturer/tradename
EDQM
mp
38-42 °C (lit.)
application(s)
pharmaceutical (small molecule)
format
neat
storage temp.
2-8°C
SMILES string
Cc1c(N)cccc1C(F)(F)F
InChI
1S/C8H8F3N/c1-5-6(8(9,10)11)3-2-4-7(5)12/h2-4H,12H2,1H3
InChI key
TWLDBACVSHADLI-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Packaging
Other Notes
related product
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
205.0 °F - closed cup
Flash Point(C)
96.1 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service