Skip to Content
Merck
All Photos(1)

Documents

W460955

Sigma-Aldrich

Guaiacol propionate

≥95%, FG

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12O3
CAS Number:
Molecular Weight:
180.20
FEMA Number:
4609
MDL number:
UNSPSC Code:
12164502
Flavis number:
09.943
NACRES:
NA.21

grade

FG

Quality Level

Agency

meets purity specifications of JECFA

reg. compliance

EU 1334/2008& 178/2002
FDA 21 CFR 117

Assay

≥95%

application(s)

flavors and fragrances

food allergen

no known allergens

Organoleptic

nutty; fruity; vanilla

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

190.0 °F

Flash Point(C)

87.78 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service