Skip to Content
Merck
All Photos(1)

Documents

T1300000

Thioridazine hydrochloride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

10-[2-(1-Methyl-2-piperidyl)ethyl]-2-(methylthio)-10H-phenothiazine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H26N2S2 · HCl
CAS Number:
Molecular Weight:
407.04
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

thioridazine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cl.CSc1ccc2Sc3ccccc3N(CCC4CCCCN4C)c2c1

InChI

1S/C21H26N2S2.ClH/c1-22-13-6-5-7-16(22)12-14-23-18-8-3-4-9-20(18)25-21-11-10-17(24-2)15-19(21)23;/h3-4,8-11,15-16H,5-7,12-14H2,1-2H3;1H

InChI key

NZFNXWQNBYZDAQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Thioridazine hydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amandeep Singh et al.
Tuberculosis (Edinburgh, Scotland), 94(6), 695-700 (2014-10-12)
Thioridazine, a potent phenothiazine compound was evaluated for its chemotherapeutic efficacy against experiment model of tuberculosis. Thioridazine potentiated the activities of both isoniazid and rifampicin (>1 log CFU reduction) against the in vitro latent Mycobacterium tuberculosis bacilli. Further, a murine
Jadel M Kratz et al.
Journal of chemical information and modeling, 54(10), 2887-2901 (2014-08-26)
The goal of this study was to design, experimentally validate, and apply a virtual screening workflow to identify novel hERG channel blockers. The hERG channel is an important antitarget in drug development since cardiotoxic risks remain as a major cause

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service