Skip to Content
Merck
All Photos(1)

Key Documents

SMB01375

Sigma-Aldrich

N-Lactoylphenylalanine

≥95% (HPLC)

Synonym(s):

(S)-N-(2-Hydroxy-1-oxopropyl)-L-phenylalanine, N-Lactoyl-Phe, N-Lactoyl-phenylalanine N-Lac-Phe, N-[(2S)-2-Hydroxy-1-oxopropyl]-L-phenylalanine, Lac-Phe

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€134.00

€134.00


Please contact Customer Service for Availability


Select a Size

Change View
10 MG
€134.00

About This Item

Empirical Formula (Hill Notation):
C12H15NO4
CAS Number:
Molecular Weight:
237.25
UNSPSC Code:
12352209
NACRES:
NA.26

€134.00


Please contact Customer Service for Availability

Product Name

N-Lactoylphenylalanine, ≥95% (HPLC)

biological source

synthetic

Quality Level

Assay

≥95% (HPLC)

form

solid

storage temp.

2-8°C

SMILES string

O=C(N[C@H](C(O)=O)CC1=CC=CC=C1)[C@@H](O)C

InChI

1S/C12H15NO4/c1-8(14)11(15)13-10(12(16)17)7-9-5-3-2-4-6-9/h2-6,8,10,14H,7H2,1H3,(H,13,15)(H,16,17)/t8-,10-/m0/s1

InChI key

IIRJJZHHNGABMQ-WPRPVWTQSA-N

General description

N-Lactoylphenylalanine, a product of metabolism that is released during exercise and subsequently suppresses appetite in mice.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An exercise-inducible metabolite that suppresses feeding and obesity
Veronica L Li, et al.
Nature, 606, 785-790 (2022)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service