Skip to Content
Merck
All Photos(1)

Key Documents

SMB00981

Sigma-Aldrich

2-Hydroxymelatonin

≥95% (HPLC)

Synonym(s):

2-OH MEL, 2-OHM, 2-OHMET, 2OHM, N-[2-(2,3-Dihydro-5-methoxy-2-oxo-1H-indol-3-yl)ethyl]acetamide, N-[2-(2-Hydroxy-5-methoxy-1H-indol-3-yl)ethyl]acetamide, AMIO

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H16N2O3
CAS Number:
Molecular Weight:
248.28
MDL number:
UNSPSC Code:
12352205
NACRES:
NA.21
Pricing and availability is not currently available.

Quality Level

Assay

≥95% (HPLC)

form

solid

storage temp.

2-8°C

SMILES string

[nH]1c2c(c(c1O)CCNC(=O)C)cc(cc2)OC

InChI key

CIEAUFSGHUWAMC-UHFFFAOYSA-N

General description

Melatonin (MT) is metabolized into several derivatives, including N1-acetyl-N2-formyl-5-methoxykynuramine (AFMK), N1-acetyl-5-methoxykynuramine (AMK), 6-hydroxymelatonin, 2-hydroxymelatonin, 3-hydroxymelatonin, and cyclic 3-hydroxymelatonin. MT and its metabolites are being studied for their antioxidant properties and their protective effects against oxidative stress in both mammals and in plants.

Application

Metabolomics research

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anis Ali Shah et al.
Chemosphere, 243, 125308-125308 (2019-11-14)
Cadmium level is continuously increasing in agricultural soils mainly due to anthropogenic activities. Cadmium is one of the most phytotoxic metals in the soils. The present study investigates the possible role of 2-hydroxymelatonin (2-OHMT) in assuagement of Cd-toxicity in cucumber
Sung Ho Lee et al.
Biochemical and biophysical research communications, 527(4), 941-946 (2020-05-23)
2-Hydroxymelatonin is a metabolite produced when melatonin 2-hydroxylase catalyzes melatonin. Recent studies have reported the important roles of melatonin in bone metabolism. However, the roles of 2-hydroxymelatonin in bone metabolism remains poorly understood. The purpose of this study is to
Kyoungwhan Back
The Plant journal : for cell and molecular biology, 105(2), 376-391 (2020-07-10)
Melatonin is a multifunctional biomolecule found in both animals and plants. In this review, the biosynthesis, levels, signaling, and possible roles of melatonin and its metabolites in plants is summarized. Tryptamine 5-hydroxylase (T5H), which catalyzes the conversion of tryptamine into
Ewy A Mathé et al.
Cancer research, 74(12), 3259-3270 (2014-04-17)
Lung cancer remains the most common cause of cancer deaths worldwide, yet there is currently a lack of diagnostic noninvasive biomarkers that could guide treatment decisions. Small molecules (<1,500 Da) were measured in urine collected from 469 patients with lung

Questions

  1. I want to know the solvent and solubility of 2-hydroxymelatonin (SMB00981).

    1 answer
    1. The solubility of this material has not been determined. However, other sources report that this compound is soluble in methanol and in 2 mM MES, containing 5 mM MgCl2, pH 5.6. Please see the links below to review example publications:
      https://www.sciencedirect.com/science/article/pii/S025462992300594X
      https://pmc.ncbi.nlm.nih.gov/articles/PMC8614918/
      https://pmc.ncbi.nlm.nih.gov/articles/PMC9219689/

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service