Skip to Content
Merck
All Photos(1)

Key Documents

SBR00026

Sigma-Aldrich

Ligstroside

Nitric oxide inhibitor, ≥95%

Synonym(s):

(-)-ligstroside, Ligustroside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H32O12
CAS Number:
Molecular Weight:
524.51
UNSPSC Code:
12352200
NACRES:
NA.25

biological source

plant

Quality Level

Assay

≥95%

form

lyophilized powder

mol wt

524.51

solubility

DMSO: ≥10 mg/mL (19 mM)
methanol: ≥10 mg/mL

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

InChI

1S/C25H32O12/c1-3-15-16(10-19(28)34-9-8-13-4-6-14(27)7-5-13)17(23(32)33-2)12-35-24(15)37-25-22(31)21(30)20(29)18(11-26)36-25/h3-7,12,16,18,20-22,24-27,29-31H,8-11H2,1-2H3/b15-3+/t16-,18+,20+,21-,22+,24-,25-/m0/s1

InChI key

GMQXOLRKJQWPNB-MVVLSVRYSA-N

General description

Ligstroside, also known as ligusroside, is a secoiridoid glycoside, a bioactive natural compound commonly derived from plants like Ligustrum lucidum, Jasminum officinale, and Osmanthus heterophyllus. Existing research suggests that this metabolite functions as an nitric oxide inhibitor and may exhibit diverse biological activities, including anti-neoplastic, antioxidant, antiviral, and anti-inflammatory properties.

Application

Ligstroside is a natural product derived from plant source that finds application in compound screening libraries, metabolomics, phytochemical, and pharmaceutical research.

Biochem/physiol Actions

Ligstroside is a natural product derived from leaves of Olea europaea. Ligstroside displays several potentially beneficial anti-inflammatory properties by exhibiting selective inhibition effect on COX-1 enzyme of arachidonate cascade metabolism. Ligstroside generates a reduction of the PGE2 (prostaglandin E2) levels in activated mouse macrophage (IC50 = 48.53 μM) and also reduction of the TXB2 (thromboxane B2) levels in human platelets (IC50 = 122.63 μM).[1] Moreover, Ligstroside exhibits inhibition activity of NO production in LPS-activated RAW264.7 macrophages (17.8% at 30 μM and 40.7% at 100 μM) without any cytotoxic effect in these cells.[2],[3] Additionally, Ligstroside exhibits in vitro antiviral activity against parainfluenza type 3 virus (IC50 = 15.6 μM).[4]

Features and Benefits

  • Suitable for Biochemical and Biomedical research
  • Versatile and adaptable for wide variety of laboratory and research applications

Storage and Stability

Hygroscopic; store in dry place with desiccant

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemical structures of constituents from the flowers of Osmanthus fragrans var. aurantiacus.
Liu, J., Nakamura, S., Xu, B., Matsumoto, T., Ohta, T., Fujimoto, K., ... & Matsuda, H.
Journal of Natural Medicines, 69(1), 135-141 (2015)
In vitro evaluation of secoiridoid glucosides from the fruits of Ligustrum lucidum as antiviral agents.
Ma, S.C., He, Z.D., Deng, X.L., But, P.P.H., Ooi, V.E.C., Xu, H.X., ... & Lee, S.F.
Chemical & Pharmaceutical Bulletin, 49(11), 1471-1473 (2001)
In vitro anti-inflammatory activity of iridoids and triterpenoid compounds isolated from Phillyrea latifolia L.
Diaz, A.M., Abad, M.J., Fernandez, L., Recuero, C., Villaescusa, L., Silvan, A.M., & Bermejo, P.
Biological & Pharmaceutical Bulletin, 23(11), 1307-1313 (2000)
In African Natural Plant Products Volume II: Discoveries and Challenges in Chemistry, Health, and Nutrition.
Bai, N., He, K., Roller, M., Lai, C.S., Shao, X., Pan, M.H., & Ho, C.T.
Journal of the American Chemical Society, 2, 115-125 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service