Skip to Content
Merck
All Photos(1)

Key Documents

S1900000

Sulfadimidine

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Sulfamethazine, 4,6-Dimethylsulfadiazine, 4-Amino-N-(4,6-dimethyl-2-pyrimidinyl)benzenesulfonamide, Sulfadimethyldiazine, Sulfadimidine

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€182.00

€182.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€182.00

About This Item

Empirical Formula (Hill Notation):
C12H14N4O2S
CAS Number:
Molecular Weight:
278.33
Beilstein:
261304
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€182.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

sulfadimidine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cc1cc(C)nc(NS(=O)(=O)c2ccc(N)cc2)n1

InChI

1S/C12H14N4O2S/c1-8-7-9(2)15-12(14-8)16-19(17,18)11-5-3-10(13)4-6-11/h3-7H,13H2,1-2H3,(H,14,15,16)

InChI key

ASWVTGNCAZCNNR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Sulfadimidine EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

An antimicrobial sulfur drug. Induces CYP3A4 expression and acetylated by N-acetyltransferase. Exhibits sex dependent pharmacokinetics, metabolized by the male specific isoform CYP2C11.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L A Poirier et al.
Regulatory toxicology and pharmacology : RTP, 30(3), 217-222 (2000-01-06)
Recently, changes have been proposed in the criteria historically used in the evaluation of the applicability to humans of some of the results obtained from the rodent carcinogenicity bioassay data. These questions center on the suitability of the rodent model
Anna Gulkowska et al.
Chemosphere, 107, 366-372 (2014-01-28)
Soil incubation experiments using (14)C-labelled sulfamethazine were carried out to assess the factors governing its nonextractable residue (NER) formation via nucleophilic addition reactions. Circumstantial evidence on possible mechanisms of NER formation was derived from a selective manipulation of soil samples.
Astrid Bienert-Zeit et al.
American journal of veterinary research, 76(4), 318-327 (2015-03-31)
To monitor concentrations of sulfadimidine in the paranasal sinus mucosa (PSM) of unsedated horses following IV administration of trimethoprim-sulfadimidine via in vivo microdialysis. 10 healthy adult horses. Concentric microdialysis probes were implanted into the subepithelial layers of the frontal sinus
Jie Hou et al.
Environmental science and pollution research international, 22(6), 4545-4554 (2014-10-17)
A feasible and rapid analysis for the simultaneous determination of sulfonamides (SAs), tetracyclines (TCs), fluoroquinolones (FQs), macrolides (MACs) and nitrofurans (NFs) in livestock manure and soils was established by solid-phase extraction (SPE)-ultra-performance liquid chromatography (UPLC)-tandem mass spectrometry (MS/MS). A total
Zhe Meng et al.
Food chemistry, 174, 597-605 (2014-12-23)
A novel UPLC-MS/MS method for quantification and confirmation of eight fluoroquinolones, five sulphonamides (SAs) and four acetyled metabolites in milk is developed. Their main fragmentation pathways were presented. The limits of quantification were in the range of 0.01-0.29 μg kg(-1)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service