Select a Size
| Pack Size | SKU | Availability | Price |
|---|---|---|---|
| 500 mL | Please contact Customer Service for Availability | €145.00 |
About This Item
grade
certified reference material
shelf life
limited shelf life, expiry date on the label
manufacturer/tradename
Paragon Scientific Ltd
viscosity
203.1 mPa.s(25.00 °C)
application(s)
cosmetics
flavors and fragrances
food and beverages
oil and petrochemicals
personal care
format
mixture
General description
1 of 1
This Item | |||
|---|---|---|---|
| format mixture | format mixture | format mixture | format mixture |
| grade certified reference material | grade certified reference material | grade certified reference material | grade certified reference material |
| application(s) cosmetics | application(s) cosmetics | application(s) cosmetics | application(s) cleaning products |
| manufacturer/tradename Paragon Scientific Ltd | manufacturer/tradename Paragon Scientific Ltd | manufacturer/tradename Paragon Scientific Ltd | manufacturer/tradename Paragon Scientific Ltd |
| viscosity 203.1 mPa.s(25.00 °C) | viscosity 300.8 mPa.s(25.00 °C) | viscosity 351.1 mPa.s(25.00 °C) | viscosity 29.04 mPa.s(25.00 °C) |
| shelf life limited shelf life, expiry date on the label | shelf life limited shelf life, expiry date on the label | shelf life limited shelf life, expiry date on the label | shelf life limited shelf life, expiry date on the label |
Still not finding the right product?
Explore all of our products under Mineral Oil Rotational Viscosity Standard
signalword
Danger
hcodes
Hazard Classifications
Asp. Tox. 1
Storage Class
10 - Combustible liquids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| RTM16-500ML | 04061837681356 |

