Skip to Content
Merck
All Photos(1)

Key Documents

RDEN1506

Relative Density Standard (15°C)

UKAS ISO/IEC17025 and ISO 17034 certified, Relative Density 15/4 0.8642 at 15 °C

Sign Into View Organizational & Contract Pricing

Select a Size

500 ML
€493.00

€493.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
500 ML
€493.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

€493.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

shelf life

limited shelf life, expiry date on the label

packaging

pkg of 500 mL

manufacturer/tradename

Paragon Scientific Ltd

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

This product is a certified reference material (CRM) certified to International Standards BS EN ISO / IEC 17025 and ISO 17034 under UKAS accreditation

Paragon ISO 17025 / ISO 17034 certified Relative Density standards are for the verification of instruments used to measure density of materials which are fluid at the desired test temperature within the range of 15°C to 150°C.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service