QCMIC005
Total & Fecal Streptococcus/Enterococcus - WP QC
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
grade
certified reference material
packaging
set of
manufacturer/tradename
RTC QC-MIC-005
application(s)
environmental
food and beverages
shipped in
wet ice
storage temp.
2-8°C
suitability
Enterococcus spp.
Streptococcus spp.
General description
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
Packaging
Set contains 2 sterile screw cap vials each containing a Vitroid (a small disk containing the organisms) and 2 - 99mL bottles of phosphate buffer.
Other Notes
This sample is designed for the quantitative analysis of fecal streptococcus. Refer to the Certificate of Analysis for additional information.
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service